Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001021.4 | NP_001012.1 | 5 | 553 | ||
NR_111943.1 | 4 | 759 | ||||
NR_111944.1 | 6 | 674 | ||||
GenBank mRNA | BC009407.2 | - | 1 | 444 | 4334552 | |
BC019899.1 | - | 1 | 449 | 3882924 | ||
BC020453.1 | - | 1 | 448 | |||
BC022370.1 | - | 5 | 466 | 4738331 | ||
BC049824.1 | - | 1 | 441 | 6212543 | ||
BC062715.1 | - | 5 | 551 | 4068842 | ||
BC070222.1 | - | 1 | 462 | 6738383 | ||
BC071928.1 | - | 1 | 452 | 5090053 | ||
BE905890.1 | - | 4 | 429 | |||
BM017772.1 | - | 5 | 555 | |||
BM457623.1 | - | 5 | 567 | |||
M13932.1 | - | 5 | 461 |