siRNA Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001021.4 | NP_001012.1 | 3 | 324 | ||
NR_111943.1 | 2 | 530 | ||||
NR_111944.1 | 3 | 324 | ||||
GenBank mRNA | AK311951.1 | - | 3 | 232 | ||
BC009407.2 | - | 1 | 215 | 4334552 | ||
BC019899.1 | - | 1 | 220 | 3882924 | ||
BC020453.1 | - | 1 | 219 | |||
BC022370.1 | - | 3 | 237 | 4738331 | ||
BC049824.1 | - | 1 | 212 | 6212543 | ||
BC062715.1 | - | 3 | 322 | 4068842 | ||
BC070222.1 | - | 1 | 233 | 6738383 | ||
BC071928.1 | - | 1 | 223 | 5090053 | ||
BE905890.1 | - | 2 | 200 | |||
BI597659.1 | - | 2 | 530 | |||
BM017772.1 | - | 3 | 325 | |||
BM457623.1 | - | 2 | 217 | |||
DA310472.1 | - | 2 | 530 | |||
M13932.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
