Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130028.1 | NP_001123500.1 | 13 | 2236 | ||
NM_003992.4 | NP_003983.2 | 13 | 1386 | |||
XM_005254151.3 | XP_005254208.1 | 13 | 2278 | |||
XM_017021906.1 | XP_016877395.1 | 13 | 2329 | |||
XM_017021907.1 | XP_016877396.1 | 13 | 2287 | |||
XM_017021909.1 | XP_016877398.1 | 18 | 6094 | |||
XM_017021910.1 | XP_016877399.1 | 16 | 5782 | |||
GenBank mRNA | AB209535.1 | - | 11 | 3222 | ||
AK026511.1 | - | 6 | 472 | |||
AK091692.1 | - | 13 | 2236 | |||
AK096115.1 | - | 8 | 2081 | |||
AK097982.1 | - | 10 | 2046 | |||
AK122860.1 | - | 9 | 3137 | |||
AK225680.1 | - | 13 | 1356 | |||
AK291983.1 | - | 13 | 1358 | |||
AL833165.1 | - | 10 | 2398 | |||
BC002555.2 | - | 13 | 1358 | 3138580 | ||
BC006103.1 | - | 12 | 1307 | 3957357 | ||
BC019881.1 | - | 13 | 1369 | 5114635 | ||
BT006993.1 | - | 12 | 1331 | |||
L29217.1 | - | 13 | 1387 | |||
L29220.1 | - | 12 | 1290 |