Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033323.3 | 8 | 1084 | |||
GenBank mRNA | AF416611.1 | - | 8 | 1041 | ||
AF420250.1 | - | 8 | 1041 | |||
AK021688.1 | - | 8 | 1041 | |||
AK292107.1 | - | 4 | 517 | |||
AK303622.1 | - | 3 | 863 | |||
AK315927.1 | - | 3 | 697 | |||
AL117583.1 | - | 1 | 829 | |||
AL833016.1 | - | 7 | 1727 | |||
BC007488.2 | - | 8 | 1138 | |||
BC082753.1 | - | 4 | 497 | 4542435 | ||
BC106049.1 | - | 4 | 565 | |||
BC107111.1 | - | 1 | 145 |
Gene Symbol: NSUN5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033322.3 | 8 | 1083 | |||
NR_104013.1 | 7 | 916 | ||||
GenBank mRNA | AF416610.1 | - | 7 | 947 | ||
AK094312.1 | - | 3 | 1859 | |||
AK126486.1 | - | 5 | 3833 | |||
AK128374.1 | - | 3 | 2071 | |||
BC107112.1 | - | 1 | 145 | 40022630 | ||
BG741008.1 | - | 5 | 625 |