Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 3 | 358 | ||
NM_001286273.1 | NP_001273202.1 | 2 | 284 | |||
NM_003295.3 | NP_003286.1 | 3 | 358 | |||
GenBank mRNA | AF072098.1 | - | 238 | |||
AK130847.1 | - | 3 | 257 | |||
AK296587.1 | - | 3 | 257 | |||
AK296952.1 | - | 2 | 784 | |||
AK312951.1 | - | 3 | 257 | |||
AY117678.1 | - | 3 | 258 | |||
AY334563.1 | - | 3 | 143 | |||
BC003352.2 | - | 3 | 241 | 2899964 | ||
BC012431.1 | - | 3 | 217 | 3880559 | ||
BC022436.1 | - | 2 | 212 | 4281364 | ||
BC040008.1 | - | 2 | 154 | |||
BC052333.1 | - | 3 | 247 | 6058799 | ||
BP215975.1 | - | 3 | 358 | |||
CR457036.1 | - | 3 | 143 | |||
X16064.1 | - | 3 | 237 | |||
X64899.1 | - | 3 | 229 |
Gene Symbol: FLJ44635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207422.2 | NP_997305.2 | 2 | 506 | ||
GenBank mRNA | AK126598.1 | - | 2 | 506 | ||
AY572224.1 | - | 2 | 506 | |||
BC137552.1 | - | 2 | 262 | 9021552 | ||
BC137564.1 | - | 2 | 262 | 9021564 |