Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021068.2 | NP_066546.1 | 1 | 594 | ||
GenBank mRNA | BC074965.2 | - | 1 | 546 | 30915507 | |
BC074966.2 | - | 1 | 546 | 30915288 | ||
BC113640.1 | - | 1 | 582 | 8322692 | ||
BC113642.1 | - | 1 | 582 | 8322694 | ||
M27318.1 | - | 1 | 666 |
Gene Symbol: IFNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002171.2 | NP_002162.1 | 1 | 572 | ||
GenBank mRNA | BC069409.1 | - | 1 | 572 | 7262186 | |
BC103972.2 | - | 1 | 572 | 40015081 | ||
BC103973.1 | - | 1 | 572 | 40015082 | ||
V00551.1 | - | 1 | 572 |