Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 149 | ||
GenBank mRNA | BC074991.2 | - | 1 | 147 | 30915521 | |
BC074992.2 | - | 1 | 147 | 30915307 | ||
BC113994.1 | - | 1 | 149 | 40084821 | ||
BC114000.1 | - | 1 | 149 | 40084820 | ||
M34913.1 | - | 1 | 149 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | E00175.1 | - | 1 | 93 |
Gene Symbol: IFNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002171.2 | NP_002162.1 | 1 | 155 | ||
GenBank mRNA | BC069409.1 | - | 1 | 155 | 7262186 | |
BC103972.2 | - | 1 | 155 | 40015081 | ||
BC103973.1 | - | 1 | 155 | 40015082 | ||
V00551.1 | - | 1 | 155 |
Gene Symbol: IFNA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002173.3 | NP_002164.1 | 1 | 115 | ||
GenBank mRNA | M28585.1 | - | 1 | 115 |
Gene Symbol: IFNA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021268.2 | NP_067091.1 | 1 | 158 | ||
GenBank mRNA | BC096732.1 | - | 1 | 151 | 40015059 | |
BC098355.1 | - | 1 | 151 | 40015056 | ||
M11026.1 | - | 1 | 158 | |||
M38289.1 | - | 1 | 251 | |||
M71246.1 | - | 1 | 70 |
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 157 | ||
GenBank mRNA | BC069329.1 | - | 1 | 117 | 7262150 | |
BC069372.1 | - | 1 | 117 | 7262162 | ||
BC069408.1 | - | 1 | 117 | 7262174 | ||
BC096699.1 | - | 1 | 151 | 40015057 | ||
BC101638.1 | - | 1 | 116 | 8069144 | ||
BC101640.1 | - | 1 | 116 | 8069146 | ||
M12350.1 | - | 1 | 157 | |||
M28586.1 | - | 1 | 157 | |||
V00540.1 | - | 1 | 117 | |||
X00145.1 | - | 1 | 157 |
Gene Symbol: IFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021068.2 | NP_066546.1 | 1 | 177 | ||
GenBank mRNA | BC074965.2 | - | 1 | 129 | 30915507 | |
BC074966.2 | - | 1 | 129 | 30915288 | ||
BC113640.1 | - | 1 | 165 | 8322692 | ||
BC113642.1 | - | 1 | 165 | 8322694 | ||
M27318.1 | - | 1 | 249 |