Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 16, 17 | 1963 | |||
GenBank mRNA | AK057241.1 | - | 14, 15 | 1441 | ||
AL833871.1 | - | 5, 6 | 443 | |||
BC144672.1 | - | 13, 14 | 1568 | |||
BC150302.1 | - | 16, 17 | 1937 | 8860399 | ||
DQ309038.1 | - | 14, 15 | 1411 |
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 16, 17 | 1975 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 16, 17 | 1963 | |||
GenBank mRNA | AK292534.1 | - | 16, 17 | 1846 | ||
BC110417.1 | - | 16, 17 | 1934 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 16, 17 | 1963 | |||
GenBank mRNA | BC063309.1 | - | 17, 18 | 1937 | ||
DQ309037.1 | - | 16, 17 | 1937 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 16, 17 | 1975 | |||
GenBank mRNA | BC066774.1 | - | 16, 17 | 1949 | ||
BC099913.1 | - | 15, 16 | 2790 | 4838463 | ||
BC125193.1 | - | 15, 16 | 1755 | 40122746 | ||
BC125194.1 | - | 15, 16 | 1755 | 40122749 |