Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022834.4 | NP_073745.2 | 2 | 831 | ||
NM_199121.2 | NP_954572.2 | 2 | 436 | |||
GenBank mRNA | AK075366.1 | - | 2 | 654 | ||
AK096773.1 | - | 2 | 243 | |||
AK291557.1 | - | 2 | 638 | |||
BC003543.1 | - | 1 | 115 | |||
BC059409.1 | - | 2 | 638 | 30349211 |