Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006083.3 | NP_006074.2 | 20 | 1771 | ||
GenBank mRNA | AF182645.1 | - | 20 | 1772 | ||
AJ005579.1 | - | 20 | 1661 | |||
AK097968.1 | - | 3 | 1798 | |||
AK225924.1 | - | 20 | 1755 | |||
AK293626.1 | - | 20 | 1599 | |||
AK312363.1 | - | 20 | 1741 | |||
AY007094.1 | - | 20 | 1694 | |||
BC013005.2 | - | 20 | 1726 | 3030079 | ||
BC051295.1 | - | 20 | 1729 | |||
BC071964.1 | - | 20 | 1719 | 6461871 |