Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004421.2 | NP_004412.2 | 13 | 1426 | ||
XM_005244731.3 | XP_005244788.1 | 13 | 1531 | |||
XM_005244732.3 | XP_005244789.1 | 13 | 1531 | |||
XM_005244733.3 | XP_005244790.1 | 13 | 1455 | |||
GenBank mRNA | AB209210.1 | - | 8 | 1449 | ||
AF006011.1 | - | 13 | 1382 | |||
AK093189.1 | - | 6 | 666 | |||
AK095867.1 | - | 6 | 644 | |||
AK293286.1 | - | 13 | 1232 | |||
AK300063.1 | - | 5 | 428 | |||
BC017225.2 | - | 11 | 1151 | |||
BC050454.1 | - | 7 | 771 | 6201003 | ||
U46461.1 | - | 13 | 1523 |