Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004421.2 | NP_004412.2 | 10 | 1050 | ||
XM_005244731.3 | XP_005244788.1 | 10 | 1080 | |||
XM_005244732.3 | XP_005244789.1 | 10 | 1080 | |||
XM_005244733.3 | XP_005244790.1 | 10 | 1079 | |||
GenBank mRNA | AB209210.1 | - | 6 | 856 | ||
AF006011.1 | - | 10 | 1006 | |||
AK093189.1 | - | 3 | 215 | |||
AK095867.1 | - | 3 | 268 | |||
AK293286.1 | - | 10 | 856 | |||
AK300063.1 | - | 2 | 52 | |||
BC017225.2 | - | 8 | 775 |