Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 6 | 860 | ||
NM_201400.3 | NP_958802.1 | 8 | 1043 | |||
NM_201598.3 | NP_963892.1 | 7 | 941 | |||
XM_005255157.4 | XP_005255214.1 | 7 | 935 | |||
XM_005255158.4 | XP_005255215.1 | 9 | 1089 | |||
XM_011522404.2 | XP_011520706.1 | 8 | 1008 | |||
GenBank mRNA | AL522535.3 | - | 6 | 782 | ||
AY037162.1 | - | 7 | 909 | |||
BC010084.2 | - | 8 | 963 | 2822323 | ||
BF314717.1 | - | 286 | ||||
BQ215173.1 | - | 2 | 107 |
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 4 | 402 | |||
GenBank mRNA | AK055771.1 | - | 2 | 1055 |
Gene Symbol: FAM86JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024250.1 | 5 | 483 | |||
NR_024251.1 | 5 | 581 | ||||
GenBank mRNA | BC032772.1 | - | 5 | 469 | ||
BC041080.1 | - | 5 | 552 | |||
BM781991.1 | - | 5 | 483 |