Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300867.1 | NP_001287796.1 | 23 | 2324 | ||
NM_001324062.1 | NP_001310991.1 | 29 | 5598 | |||
NM_001324063.1 | NP_001310992.1 | 26 | 5349 | |||
NM_001324064.1 | NP_001310993.1 | 29 | 5571 | |||
NM_001324065.1 | NP_001310994.1 | 24 | 2396 | |||
NM_198551.3 | NP_940953.2 | 28 | 5526 | |||
XM_006711304.3 | XP_006711367.2 | 27 | 5733 | |||
XM_017001243.1 | XP_016856732.1 | 29 | 5423 | |||
GenBank mRNA | AK025122.1 | - | 20 | 1825 | ||
AK025506.1 | - | 25 | 3206 | |||
AK026489.1 | - | 17 | 1585 | |||
AK290335.1 | - | 19 | 1801 | |||
AK296712.1 | - | 23 | 2305 | |||
AX775845.1 | - | 23 | 2298 | |||
BC047116.1 | - | 13 | 1198 | |||
D87742.1 | - | 25 | 3375 |