Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016582.2 | NP_057666.1 | 7, 8 | 1826 | ||
NR_027391.1 | 6, 7 | 1657 | ||||
GenBank mRNA | AB020598.1 | - | 7, 8 | 1826 | ||
AF135600.1 | - | 7, 8 | 1719 | |||
AK127216.1 | - | 2, 3 | 4281 | |||
BC025710.1 | - | 6, 7 | 1517 | 5224878 | ||
BC037974.1 | - | 7, 8 | 1686 | 5213632 |