Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213606.3 | NP_998771.3 | 8 | 3795 | ||
XM_017016237.1 | XP_016871726.1 | 7 | 3885 | |||
XM_017016238.1 | XP_016871727.1 | 7 | 3824 | |||
XM_017016239.1 | XP_016871728.1 | 7 | 3683 | |||
GenBank mRNA | AK124901.1 | - | 4 | 3075 | ||
AK127303.1 | - | 8 | 3796 | |||
BC086873.1 | - | 3 | 2749 | |||
CR936657.1 | - | 6 | 5993 |
Gene Symbol: AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000476.2 | NP_000467.1 | 7 | 1612 | ||
NM_001318121.1 | NP_001305050.1 | 7 | 1599 | |||
NM_001318122.1 | NP_001305051.1 | 6 | 1607 | |||
GenBank mRNA | EU794628.1 | - | 7 | 1612 |
Gene Symbol: BRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024482.1 | - | 9 | 2534 | ||
AK131083.1 | - | 8 | 3733 | |||
AK131097.1 | - | 2 | 3898 |
Gene Symbol: CRYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC039589.1 | - | 1 | 912 | ||
BX647276.1 | - | 11 | 1814 | |||
BX648547.1 | - | 13 | 1948 |
Gene Symbol: RNF115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272952.4 | XP_005273009.1 | 10 | 2498 |
Gene Symbol: SSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292008.1 | NP_001278937.1 | 8 | 7512 | ||
NM_003144.4 | NP_003135.2 | 8 | 7716 | |||
NR_120448.1 | 7 | 7637 | ||||
GenBank mRNA | AK024294.1 | - | 1 | 3051 |