Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC022796.1 | - | 3 | 798 | 4811294 | |
BC069526.1 | - | 3 | 465 | 7262120 | ||
BC106059.1 | - | 3 | 806 | 4992804 | ||
BX334905.2 | - | 3 | 864 |
Gene Symbol: CGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033043.1 | NP_149032.1 | 3 | 821 | ||
GenBank mRNA | BC006290.2 | - | 3 | 782 | 3960125 | |
BC051378.1 | - | 3 | 782 | 3956670 | ||
BC103970.1 | - | 3 | 552 | 40014677 | ||
BC103971.1 | - | 3 | 548 | 40014679 | ||
BC106723.1 | - | 3 | 466 | 40014469 | ||
BC106724.1 | - | 3 | 465 | 40014470 | ||
BT006890.1 | - | 2 | 456 |
Gene Symbol: CGB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033142.1 | NP_149133.1 | 3 | 821 | ||
XM_017027502.1 | XP_016882991.1 | 6 | 2168 | |||
GenBank mRNA | AK092722.1 | - | 5 | 1891 | ||
AK125108.1 | - | 5 | 3328 |
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 3 | 828 | ||
GenBank mRNA | BC030994.1 | - | 3 | 823 | 4605021 | |
BC103969.1 | - | 3 | 548 | 40014676 | ||
BC128603.1 | - | 3 | 548 | 40002893 |