Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251994.3 | XP_005252051.1 | 16 | 5736 | ||
XM_005251995.3 | XP_005252052.1 | 15 | 5711 | |||
XM_005251996.3 | XP_005252053.1 | 14 | 5680 | |||
XM_011518708.2 | XP_011517010.1 | 7 | 3869 | |||
XM_017014734.1 | XP_016870223.1 | 15 | 5472 |
Gene Symbol: ADAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131102.1 | - | 1 | 1383 |
Gene Symbol: ANKRD36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB353103.1 | - | 1 | 191 |
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648159.1 | - | 1 | 1273 |
Gene Symbol: AP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209422.1 | - | 4 | 2158 |
Gene Symbol: C12orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056213.1 | - | 4 | 2216 |
Gene Symbol: C17orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125817.1 | - | 6 | 3140 |
Gene Symbol: CDC42BPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126684.1 | - | 1 | 1691 |
Gene Symbol: CDK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300960.1 | NP_001287889.1 | 12 | 4348 | ||
NM_001300963.1 | NP_001287892.1 | 14 | 4296 | |||
NM_001300964.1 | NP_001287893.1 | 13 | 4250 | |||
NM_015076.4 | NP_055891.1 | 13 | 4480 | |||
XM_005266871.3 | XP_005266928.1 | 10 | 4063 | |||
XM_011535630.2 | XP_011533932.1 | 13 | 4826 | |||
XM_011535631.2 | XP_011533933.1 | 12 | 4698 | |||
XM_017010587.1 | XP_016866076.1 | 14 | 4706 | |||
XM_017010588.1 | XP_016866077.1 | 11 | 4172 | |||
GenBank mRNA | AB028951.2 | - | 13 | 4297 | ||
AL122055.1 | - | 10 | 3933 |
Gene Symbol: CERS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA230084.1 | - | 1 | 256 |
Gene Symbol: CFAP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024878.1 | - | 1 | 1117 |
Gene Symbol: CTSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648239.1 | - | 1 | 2072 |
Gene Symbol: DAW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127882.1 | - | 11 | 1648 |
Gene Symbol: DBNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209486.1 | - | 11 | 3506 |
Gene Symbol: DCAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA634196.1 | - | 1 | 47 |
Gene Symbol: DIDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127121.1 | - | 1 | 211 |
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB960096.1 | - | 4 | 356 |
Gene Symbol: DRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019578.1 | XP_016875067.1 | 2 | 633 |
Gene Symbol: EHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092866.1 | - | 1 | 1007 |
Gene Symbol: EMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271427.1 | NP_001258356.1 | 23 | 4857 | ||
NM_001271428.1 | NP_001258357.1 | 23 | 4857 | |||
NM_001271429.1 | NP_001258358.1 | 22 | 4794 | |||
NM_015047.2 | NP_055862.1 | 23 | 4860 | |||
GenBank mRNA | AK124740.1 | - | 3 | 3297 | ||
D42044.2 | - | 23 | 4810 |
Gene Symbol: FAM20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324310.1 | NP_001311239.1 | 8 | 2649 | ||
NM_001324311.1 | NP_001311240.1 | 8 | 2659 | |||
NM_014864.3 | NP_055679.1 | 8 | 2754 | |||
XM_017003001.1 | XP_016858490.1 | 9 | 2700 | |||
XM_017003002.1 | XP_016858491.1 | 9 | 2693 | |||
GenBank mRNA | AB007944.1 | - | 8 | 2796 | ||
AB480690.1 | - | 8 | 2754 | |||
BC051794.1 | - | 8 | 2754 | 5492043 |
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127774.1 | - | 3 | 326 |
Gene Symbol: FLJ45508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127416.1 | - | 1 | 1603 |
Gene Symbol: GCDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102316.1 | 5 | 414 | |||
GenBank mRNA | AK296446.1 | - | 5 | 377 |
Gene Symbol: GRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK300225.1 | - | 7 | 1399 |
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094412.1 | - | 1 | 1564 |
Gene Symbol: GUCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284251.1 | NP_001271180.1 | 6 | 2420 | ||
NM_001284252.1 | NP_001271181.1 | 6 | 2417 | |||
NM_001284253.1 | NP_001271182.1 | 6 | 2288 | |||
NM_001284254.1 | NP_001271183.1 | 6 | 2455 | |||
NM_001284255.1 | NP_001271184.1 | 6 | 2326 | |||
NM_001284256.1 | NP_001271185.1 | 5 | 2216 | |||
NM_001284257.1 | NP_001271186.1 | 5 | 2213 | |||
NM_031444.3 | NP_113632.2 | 6 | 2458 | |||
NR_104286.1 | 4 | 2093 | ||||
XM_005261761.1 | XP_005261818.1 | 5 | 2176 | |||
GenBank mRNA | AK024723.1 | - | 1 | 170 | ||
AK025242.1 | - | 1 | 1320 | |||
BC002924.2 | - | 5 | 2113 | |||
BC053529.1 | - | 6 | 2112 | |||
BC070109.1 | - | 6 | 2461 | 4838028 |
Gene Symbol: KLHL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB062478.1 | - | 1 | 43 |
Gene Symbol: LIPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713529.3 | XP_006713592.1 | 9 | 3383 | ||
XM_011512530.2 | XP_011510832.1 | 11 | 3650 | |||
XM_011512531.2 | XP_011510833.1 | 12 | 3595 | |||
XM_017005852.1 | XP_016861341.1 | 9 | 3371 |
Gene Symbol: LOC101926955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110003.1 | 4 | 3583 |
Gene Symbol: LOC283861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098143.1 | - | 1 | 1437 |
Gene Symbol: LOC399715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040079.2 | 6 | 2276 | |||
GenBank mRNA | AK128185.1 | - | 7 | 2394 |
Gene Symbol: LOC400576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127731.1 | - | 2 | 2341 |
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205138.3 | NP_001192067.1 | 5 | 1348 | ||
NM_001271471.2 | NP_001258400.1 | 5 | 1767 | |||
NM_001318803.1 | NP_001305732.1 | 6 | 1442 | |||
NM_145309.5 | NP_660352.1 | 6 | 1485 | |||
NR_026886.3 | 7 | 1665 | ||||
NR_134858.1 | 4 | 1264 | ||||
XM_006718473.3 | XP_006718536.1 | 6 | 1711 | |||
XM_006718474.3 | XP_006718537.1 | 5 | 1750 | |||
XM_011544847.2 | XP_011543149.1 | 6 | 1939 | |||
XM_011544848.2 | XP_011543150.1 | 7 | 1997 | |||
XM_017017359.1 | XP_016872848.1 | 7 | 2102 | |||
XM_017017360.1 | XP_016872849.1 | 7 | 1954 | |||
GenBank mRNA | AK298997.1 | - | 5 | 866 | ||
BC012855.1 | - | 6 | 957 | 3863323 | ||
EU627066.1 | - | 6 | 957 | |||
EU627068.1 | - | 5 | 1238 |
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647201.1 | - | 1 | 2231 |
Gene Symbol: MORC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028311.1 | XP_016883800.1 | 1 | 34 |
Gene Symbol: MSH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209886.1 | - | 2 | 3473 |
Gene Symbol: NFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC018471.1 | - | 5 | 463 |
Gene Symbol: NR1H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091535.1 | - | 4 | 327 |
Gene Symbol: NTAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092915.1 | - | 1 | 977 |
Gene Symbol: PAPOLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252006.1 | NP_001238935.1 | 9 | 2018 | ||
NM_001252007.1 | NP_001238936.1 | 7 | 2852 |
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006715228.2 | XP_006715291.1 | 6 | 2856 | ||
XM_011514942.2 | XP_011513244.1 | 6 | 2837 | |||
XM_011514943.2 | XP_011513245.1 | 6 | 2822 | |||
XM_011514944.2 | XP_011513246.1 | 6 | 3441 | |||
XM_017011361.1 | XP_016866850.1 | 6 | 2863 | |||
XM_017011362.1 | XP_016866851.1 | 7 | 2972 | |||
XM_017011363.1 | XP_016866852.1 | 7 | 3740 |
Gene Symbol: PSPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020650.1 | XP_016876139.1 | 7 | 2040 |
Gene Symbol: RNF115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HY099262.1 | - | 1 | 352 |
Gene Symbol: RPS15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023088.1 | - | 4 | 256 |
Gene Symbol: SAMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091238.1 | - | 1 | 1242 |
Gene Symbol: SCARB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209436.1 | - | 11 | 4106 |
Gene Symbol: SCN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199037.4 | NP_950238.1 | 3 | 3652 |
Gene Symbol: SIX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017420.4 | NP_059116.3 | 3 | 4070 |
Gene Symbol: SLFN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104587.1 | NP_001098057.1 | 3 | 423 | ||
NM_001104588.1 | NP_001098058.1 | 3 | 341 | |||
NM_001104589.1 | NP_001098059.1 | 2 | 263 | |||
NM_001104590.1 | NP_001098060.1 | 3 | 253 | |||
XM_006722171.3 | XP_006722234.1 | 3 | 234 | |||
XM_011525465.2 | XP_011523767.1 | 2 | 456 | |||
XM_017025299.1 | XP_016880788.1 | 3 | 574 | |||
GenBank mRNA | AK092241.1 | - | 3 | 176 | ||
BP245452.1 | - | 3 | 341 | |||
BX490030.1 | - | 3 | 342 | |||
CB265500.1 | - | 2 | 169 |
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316897.1 | NP_001303826.1 | 4 | 1685 | ||
NM_001316898.1 | NP_001303827.1 | 4 | 1678 | |||
NR_133631.1 | 3 | 1475 | ||||
GenBank mRNA | AK075259.1 | - | 4 | 1679 | ||
AK130389.1 | - | 1 | 1759 | |||
BC012940.1 | - | 4 | 1635 | 4478288 |
Gene Symbol: SPATA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145207.2 | NP_660208.2 | 16 | 3506 |
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127482.2 | NP_001120954.1 | 4 | 2425 | ||
NM_020456.3 | NP_065189.1 | 5 | 2542 | |||
NR_023351.2 | 3 | 2258 | ||||
GenBank mRNA | AF334405.1 | - | 5 | 2381 |
Gene Symbol: STRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723174.2 | XP_006723237.1 | 1 | 39 | ||
GenBank mRNA | AK298804.1 | - | 2 | 139 |
Gene Symbol: SYTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267215.4 | XP_005267272.1 | 18 | 3382 | ||
XM_005267218.4 | XP_005267275.1 | 18 | 2693 | |||
XM_005267222.4 | XP_005267279.1 | 10 | 1964 | |||
XM_006715605.3 | XP_006715668.1 | 19 | 3474 | |||
XM_006715606.3 | XP_006715669.1 | 19 | 3395 | |||
XM_006715611.3 | XP_006715674.1 | 18 | 2997 | |||
XM_011536254.2 | XP_011534556.1 | 19 | 3163 | |||
XM_011536255.2 | XP_011534557.1 | 9 | 2219 | |||
XM_017011496.1 | XP_016866985.1 | 18 | 3563 | |||
XM_017011497.1 | XP_016866986.1 | 19 | 2900 | |||
XM_017011498.1 | XP_016866987.1 | 15 | 2713 |
Gene Symbol: THSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358149.1 | - | 5 | 1427 |
Gene Symbol: TMEM116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020192.1 | XP_016875681.1 | 3 | 1605 |
Gene Symbol: TROAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537725.2 | XP_011536027.1 | 5 | 1380 |
Gene Symbol: TTC21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024753.4 | NP_079029.3 | 29 | 4535 | ||
XM_011511871.2 | XP_011510173.1 | 24 | 7780 | |||
XM_011511872.2 | XP_011510174.1 | 29 | 4509 | |||
XM_017004968.1 | XP_016860457.1 | 24 | 6026 | |||
XM_017004969.1 | XP_016860458.1 | 27 | 4195 | |||
GenBank mRNA | BC035767.2 | - | 6 | 1301 | ||
BX647903.1 | - | 15 | 2286 |
Gene Symbol: UBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309258.1 | - | 3 | 855 |
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080435.2 | NP_001073904.1 | 10 | 3090 | ||
XM_005272421.4 | XP_005272478.1 | 10 | 2706 | |||
XM_005272422.3 | XP_005272479.1 | 10 | 2453 | |||
XM_005272423.4 | XP_005272480.1 | 10 | 2692 | |||
XM_011521232.2 | XP_011519534.1 | 8 | 2902 | |||
GenBank mRNA | AB075851.1 | - | 10 | 3076 | ||
AK097947.1 | - | 2 | 1214 | |||
AK126887.1 | - | 10 | 3092 |
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022098.3 | NP_071381.1 | 10 | 5379 | ||
GenBank mRNA | BX648018.1 | - | 11 | 3235 |
Gene Symbol: ZNF264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003417.4 | NP_003408.1 | 4 | 3720 | ||
XM_011527522.2 | XP_011525824.1 | 3 | 3624 | |||
GenBank mRNA | AB007872.1 | - | 5 | 5224 |
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023889.1 | XP_016879378.1 | 7 | 2649 |
Gene Symbol: ZNF674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039891.2 | NP_001034980.1 | 6 | 2962 | ||
NM_001146291.1 | NP_001139763.1 | 6 | 2944 | |||
NM_001190417.1 | NP_001177346.1 | 6 | 2947 |
Gene Symbol: ZNF788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027049.1 | 3 | 3022 | |||
GenBank mRNA | AK128282.1 | - | 1 | 2079 |