Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130013.1 | - | 3 | 1971 |
Gene Symbol: ADRA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648936.1 | - | 1 | 2905 |
Gene Symbol: ATXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S75313.1 | - | 10 | 1282 |
Gene Symbol: C12orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143905.2 | NP_001137377.1 | 3 | 909 | ||
NM_001194995.1 | NP_001181924.1 | 3 | 1202 | |||
NM_152269.4 | NP_689482.1 | 3 | 993 | |||
XM_005253630.4 | XP_005253687.1 | 4 | 1013 | |||
XM_011538980.2 | XP_011537282.1 | 3 | 1771 | |||
GenBank mRNA | AF061733.1 | - | 3 | 834 | ||
AK095982.1 | - | 3 | 1373 | |||
BC018145.1 | - | 3 | 794 | 3846678 | ||
BC062329.1 | - | 3 | 836 | 5484911 |
Gene Symbol: C20orf196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529179.2 | XP_011527481.1 | 3 | 2360 | ||
XM_011529181.2 | XP_011527483.1 | 3 | 2358 |
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028676.1 | - | 1 | 1190 |
Gene Symbol: DKFZp434E1119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL834516.1 | - | 1 | 3360 |
Gene Symbol: DNAJC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181706.4 | NP_859057.4 | 5 | 1722 | ||
XM_017017216.1 | XP_016872705.1 | 5 | 2495 | |||
GenBank mRNA | AL833128.1 | - | 5 | 1712 |
Gene Symbol: GAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007966.1 | XP_016863455.1 | 2 | 6972 | ||
XM_017007968.1 | XP_016863457.1 | 2 | 6970 |
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI921211.1 | - | 1 | 67 | ||
DB299262.1 | - | 1 | 417 |
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273120.2 | XP_005273177.1 | 3 | 2871 | ||
GenBank mRNA | BC015009.1 | - | 3 | 2719 | 4862726 |
Gene Symbol: KCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270851.4 | XP_005270908.1 | 8 | 4497 | ||
XM_006710629.3 | XP_006710692.1 | 8 | 4807 | |||
XM_006710630.3 | XP_006710693.1 | 7 | 4440 | |||
XM_017001244.1 | XP_016856733.1 | 8 | 4822 |
Gene Symbol: KIF13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514806.2 | XP_011513108.1 | 3 | 328 |
Gene Symbol: LINC01010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055166.1 | - | 3 | 1987 |
Gene Symbol: LOC100287846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037168.1 | 1 | 307 | |||
GenBank mRNA | DB469678.1 | - | 1 | 308 |
Gene Symbol: LOC101927282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110753.1 | 3 | 209 | |||
GenBank mRNA | BC035172.1 | - | 3 | 216 |
Gene Symbol: LOC105371849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135648.1 | 2 | 572 |
Gene Symbol: LOC105374956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098665.1 | - | 8 | 1486 |
Gene Symbol: LOC399851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY129010.1 | - | 2 | 1923 |
Gene Symbol: LOC646471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024498.1 | 1 | 809 |
Gene Symbol: MAFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161572.1 | NP_001155044.1 | 3 | 2076 | ||
NM_001161573.1 | NP_001155045.1 | 3 | 1834 | |||
NM_001161574.1 | NP_001155046.1 | 2 | 1983 | |||
NM_012323.3 | NP_036455.1 | 3 | 2050 | |||
GenBank mRNA | AB025247.1 | - | 3 | 1884 | ||
AJ010857.1 | - | 3 | 1966 | |||
AK002001.1 | - | 3 | 1991 | |||
AK097953.1 | - | 3 | 1941 | |||
BC015037.1 | - | 3 | 1923 | 3922552 | ||
BC067751.1 | - | 3 | 1989 | 30335753 |
Gene Symbol: MYLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308301.1 | NP_001295230.1 | 12 | 5110 | ||
NM_182493.2 | NP_872299.2 | 13 | 5929 |
Gene Symbol: NUP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022106.1 | - | 1 | 2656 |
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749310.1 | - | 5 | 495 |
Gene Symbol: ORAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126340.2 | NP_001119812.1 | 4 | 4913 | ||
NM_001271818.1 | NP_001258747.1 | 4 | 4936 | |||
NM_001271819.1 | NP_001258748.1 | 3 | 4675 | |||
NM_032831.3 | NP_116220.1 | 3 | 4804 |
Gene Symbol: PHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832725.1 | - | 9 | 1469 |
Gene Symbol: PROKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529159.2 | XP_011527461.1 | 3 | 4261 | ||
XM_017027646.1 | XP_016883135.1 | 3 | 3545 |
Gene Symbol: PTPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019724.1 | XP_016875213.1 | 1 | 60 | ||
GenBank mRNA | BC051329.1 | - | 10 | 2395 |
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243764.1 | NP_001230693.1 | 7 | 3124 | ||
NM_001243765.1 | NP_001230694.1 | 9 | 3732 | |||
XM_005245680.1 | XP_005245737.1 | 26 | 5546 | |||
XM_005245681.2 | XP_005245738.1 | 26 | 5505 | |||
XM_006711693.1 | XP_006711756.1 | 25 | 5348 | |||
XM_011510223.2 | XP_011508525.1 | 27 | 5573 | |||
XM_017002995.1 | XP_016858484.1 | 10 | 4818 | |||
XM_017002996.1 | XP_016858485.1 | 9 | 3301 | |||
XM_017002997.1 | XP_016858486.1 | 9 | 3409 | |||
XM_017002998.1 | XP_016858487.1 | 10 | 10396 | |||
XM_017002999.1 | XP_016858488.1 | 9 | 3209 | |||
GenBank mRNA | AB007940.1 | - | 10 | 3711 | ||
CR627374.1 | - | 8 | 3164 |
Gene Symbol: RFPL1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002727.2 | 1 | 3984 | |||
GenBank mRNA | AJ010230.1 | - | LIT2045 RFPL1S |
1 | 3984 |
Gene Symbol: SHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094391.1 | - | 6 | 1981 | ||
AK123658.1 | - | 1 | 1746 |
Gene Symbol: SOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000636.3 | NP_000627.2 | 5 | 3424 | ||
NM_001322814.1 | NP_001309743.1 | 4 | 3307 | |||
NM_001322815.1 | NP_001309744.1 | 4 | 3244 | |||
NM_001322819.1 | NP_001309748.1 | 5 | 3330 | |||
NM_001322820.1 | NP_001309749.1 | 5 | 3333 | |||
GenBank mRNA | BC001980.1 | - | 1 | 463 |
Gene Symbol: SPARCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291976.1 | NP_001278905.1 | 3 | 610 | ||
GenBank mRNA | AK316533.1 | - | 3 | 200 |
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 18 | 4115 | ||
XM_011518446.2 | XP_011516748.1 | 18 | 4130 | |||
XM_011518447.1 | XP_011516749.1 | 17 | 4091 | |||
XM_011518449.1 | XP_011516751.1 | 17 | 3998 | |||
XM_011518451.1 | XP_011516753.1 | 16 | 3986 | |||
XM_011518452.1 | XP_011516754.1 | 17 | 3881 | |||
XM_017014571.1 | XP_016870060.1 | 17 | 4088 | |||
XM_017014572.1 | XP_016870061.1 | 16 | 3857 | |||
GenBank mRNA | AF153415.1 | - | 18 | 4119 | ||
AK074248.1 | - | 8 | 2413 | |||
AK090543.1 | - | 4 | 3170 | |||
BC014248.2 | - | 8 | 2466 |
Gene Symbol: TSPAN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016010.1 | XP_016871499.1 | 9 | 4750 |
Gene Symbol: TSPAN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278740.1 | NP_001265669.1 | 8 | 3135 | ||
NM_001278741.1 | NP_001265670.1 | 8 | 2905 | |||
NM_001278742.1 | NP_001265671.1 | 7 | 2797 | |||
NM_001278743.1 | NP_001265672.1 | 7 | 2821 | |||
NM_003270.3 | NP_003261.1 | 8 | 2843 |
Gene Symbol: YME1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253866.1 | NP_001240795.1 | 18 | 2773 | ||
NM_014263.3 | NP_055078.1 | 19 | 2872 | |||
NM_139312.2 | NP_647473.1 | 20 | 3043 | |||
XM_011519300.2 | XP_011517602.1 | 19 | 3326 | |||
GenBank mRNA | AF151782.1 | - | 20 | 2887 | ||
AJ132637.1 | - | 19 | 2846 | |||
AK001259.1 | - | 10 | 1632 | |||
BC023507.2 | - | 19 | 2814 | 3637576 |
Gene Symbol: ZNF346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308214.1 | NP_001295143.1 | 6 | 4099 | ||
NM_001308215.1 | NP_001295144.1 | 6 | 4009 | |||
NM_001308216.1 | NP_001295145.1 | 4 | 3770 | |||
NM_001308218.1 | NP_001295147.1 | 4 | 3901 | |||
NM_001308221.1 | NP_001295150.1 | 3 | 3667 | |||
NM_012279.3 | NP_036411.1 | 7 | 4195 | |||
NR_131773.1 | 6 | 4050 | ||||
NR_131774.1 | 5 | 3864 | ||||
XM_011534495.2 | XP_011532797.1 | 8 | 3670 | |||
GenBank mRNA | DB548047.1 | - | 1 | 341 |
Gene Symbol: ZNF589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048798.1 | - | 5 | 4404 |
Gene Symbol: ZNRF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046851.1 | 3 | 1915 | |||
GenBank mRNA | AK095488.1 | - | 3 | 1912 |
Gene Symbol: ZZEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122719.1 | - | 15 | 3136 |