Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI832214.1 | - | 2 | 320 |
Gene Symbol: ACSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024560.2 | NP_078836.1 | 16 | 2820 | ||
XM_005269150.2 | XP_005269207.1 | 16 | 2754 | |||
XM_011538740.2 | XP_011537042.1 | 16 | 2469 | |||
GenBank mRNA | AK074267.1 | - | 6 | 1244 | ||
AK074938.1 | - | 11 | 2001 |
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005248137.4 | XP_005248194.1 | 17 | 3467 | ||
GenBank mRNA | BC020926.1 | - | 1 | 1899 |
Gene Symbol: AKAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK303640.1 | - | 8 | 1223 |
Gene Symbol: BISPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130765.1 | 5 | 872 | |||
NR_130766.1 | 4 | 764 | ||||
GenBank mRNA | AK311380.1 | - | 4 | 981 |
Gene Symbol: BOLA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045634.1 | 2 | 2066 | |||
NR_045636.1 | 3 | 1940 |
Gene Symbol: C19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526460.2 | XP_011524762.1 | 10 | 1199 |
Gene Symbol: CCDC142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032779.3 | NP_116168.3 | 9 | 3152 | ||
GenBank mRNA | AK027303.1 | - | 3 | 1350 |
Gene Symbol: CEP89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001375.1 | - | 2 | 1000 | ||
BC029978.1 | - | 2 | 964 |
Gene Symbol: CHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094363.1 | - | 1 | 2478 | ||
BX391712.2 | - | 1 | 177 |
Gene Symbol: CPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005502.2 | NP_001005502.1 | 9 | 5003 | ||
NM_001874.4 | NP_001865.1 | 9 | 5035 | |||
NM_198320.3 | NP_938079.1 | 9 | 5021 |
Gene Symbol: DENR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG705827.1 | - | 4 | 595 |
Gene Symbol: DEPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013735.1 | XP_016869224.1 | 7 | 3608 |
Gene Symbol: DPY19L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC050303.1 | - | 1 | 2232 |
Gene Symbol: F11R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016946.4 | NP_058642.1 | 10 | 1912 | ||
GenBank mRNA | AF154005.1 | - | 10 | 1584 | ||
AF172398.2 | - | 10 | 1788 | |||
AF207907.1 | - | 10 | 1657 | |||
AK075152.1 | - | 10 | 1712 | |||
AY358896.1 | - | 10 | 1693 | |||
BC001533.2 | - | 10 | 1686 | 2990019 |
Gene Symbol: FAM53B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120630.1 | 3 | 1542 | |||
NR_120631.1 | 2 | 1402 | ||||
GenBank mRNA | BC041007.2 | - | 2 | 1402 |
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024963.5 | NP_079239.3 | 5 | 6922 | ||
GenBank mRNA | AK123535.1 | - | 2 | 3101 |
Gene Symbol: FBXO24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310487.1 | - | 4 | 925 |
Gene Symbol: FBXO27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178820.4 | NP_849142.1 | 6 | 1612 | ||
GenBank mRNA | AF436061.1 | - | 6 | 1555 | ||
BC014527.2 | - | 6 | 1574 | 3841901 | ||
BC030060.1 | - | 6 | 1541 | 5421414 |
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308028.1 | NP_001294957.1 | 18 | 3212 | ||
NM_001308031.1 | NP_001294960.1 | 11 | 2162 | |||
NM_005246.3 | NP_005237.2 | 20 | 3598 | |||
XM_011543276.2 | XP_011541578.1 | 12 | 2414 |
Gene Symbol: FN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641150.1 | - | 7 | 4133 |
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092106.1 | - | 1 | 1742 |
Gene Symbol: GPR142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257305.3 | XP_005257362.1 | 4 | 2191 |
Gene Symbol: GTF2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057670.1 | - | 1 | 1125 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X83299.1 | - | 7 | 1067 |
Gene Symbol: HEXDC-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129961.1 | - | 1 | 1262 |
Gene Symbol: ICA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288622.1 | NP_001275551.1 | 13 | 3229 | ||
NM_001288623.1 | NP_001275552.1 | 13 | 3174 | |||
NM_138468.5 | NP_612477.3 | 14 | 3356 | |||
GenBank mRNA | AK055978.1 | - | 13 | 2971 | ||
AK092310.1 | - | 13 | 3229 | |||
AL833904.1 | - | 14 | 3013 |
Gene Symbol: IL12RB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270825.2 | XP_005270882.1 | 17 | 4144 | ||
XM_005270827.2 | XP_005270884.1 | 17 | 4140 | |||
XM_005270828.3 | XP_005270885.1 | 16 | 4052 | |||
XM_006710617.2 | XP_006710680.1 | 15 | 3886 | |||
XM_011541383.2 | XP_011539685.1 | 17 | 3596 | |||
XM_011541384.2 | XP_011539686.1 | 16 | 3985 | |||
XM_017001203.1 | XP_016856692.1 | 15 | 3956 |
Gene Symbol: INPP5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833055.1 | - | 6 | 3873 |
Gene Symbol: KLHL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037989.1 | - | 3 | 468 |
Gene Symbol: LINC00407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG959100.1 | - | 4 | 376 |
Gene Symbol: LINC01547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127059.1 | - | 1 | 1571 |
Gene Symbol: LOC101927666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110809.1 | 3 | 4107 | |||
GenBank mRNA | CR936603.1 | - | 3 | 4107 |
Gene Symbol: LOC105371022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093043.1 | - | 2 | 780 |
Gene Symbol: LOC401320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096687.1 | - | 3 | 2556 |
Gene Symbol: LOC619427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL834492.1 | - | 1 | 2893 |
Gene Symbol: LOC730102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092831.1 | - | 2 | 2324 |
Gene Symbol: LRRC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097393.1 | - | 2 | 278 |
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181725.3 | NP_859076.3 | 9 | 2279 | ||
GenBank mRNA | AK000991.1 | - | 4 | 1525 | ||
AK022822.1 | - | 9 | 2258 |
Gene Symbol: NEK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534040.2 | XP_011532342.1 | 13 | 2597 |
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308426.1 | NP_001295355.1 | 6 | 2741 | ||
NM_001308427.1 | NP_001295356.1 | 6 | 2550 | |||
NM_001308428.1 | NP_001295357.1 | 6 | 2541 | |||
NM_001308430.1 | NP_001295359.1 | 5 | 2484 | |||
NM_001308431.1 | NP_001295360.1 | 5 | 2701 | |||
NM_001308433.1 | NP_001295362.1 | 5 | 2501 | |||
NM_001308434.1 | NP_001295363.1 | 5 | 2644 | |||
NM_001308435.1 | NP_001295364.1 | 4 | 2540 | |||
NM_005793.4 | NP_005784.1 | 6 | 2524 | |||
XM_017005512.1 | XP_016861001.1 | 3 | 3075 | |||
XM_017005513.1 | XP_016861002.1 | 6 | 2466 | |||
XM_017005514.1 | XP_016861003.1 | 7 | 2723 | |||
XM_017005515.1 | XP_016861004.1 | 5 | 2479 | |||
XM_017005517.1 | XP_016861006.1 | 3 | 2241 | |||
XM_017005518.1 | XP_016861007.1 | 4 | 2370 | |||
GenBank mRNA | DA912668.1 | - | 1 | 70 |
Gene Symbol: NUMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK226069.1 | - | 1 | 1240 |
Gene Symbol: PCAT29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097902.1 | - | 2 | 1892 |
Gene Symbol: PHLDB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021690.1 | - | 2 | 1153 |
Gene Symbol: PLEKHH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124381.1 | - | 7 | 2217 | ||
AL833546.1 | - | 15 | 3667 |
Gene Symbol: PLXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020405.4 | NP_065138.2 | 14 | 2724 | ||
GenBank mRNA | AF378753.1 | - | 14 | 2594 | ||
AL833517.1 | - | 6 | 1600 |
Gene Symbol: POLR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ653001.1 | - | 1 | 492 |
Gene Symbol: PSPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW574794.1 | - | 1 | 175 |
Gene Symbol: RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF217985.1 | - | 1 | 473 |
Gene Symbol: RASSF8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055961.1 | - | 1 | 1392 |
Gene Symbol: RBPMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006867.3 | NP_006858.1 | 8 | 1348 | ||
XM_017012980.1 | XP_016868469.1 | 9 | 1480 | |||
XM_017012983.1 | XP_016868472.1 | 9 | 2466 | |||
XM_017012985.1 | XP_016868474.1 | 6 | 909 | |||
XM_017012990.1 | XP_016868479.1 | 7 | 1212 | |||
XM_017012991.1 | XP_016868480.1 | 10 | 1627 | |||
GenBank mRNA | D84110.1 | - | 8 | 1322 | ||
N88549.1 | - | 1 | 103 |
Gene Symbol: RNF125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000463.1 | - | 6 | 1663 | ||
AK225518.1 | - | 6 | 1663 |
Gene Symbol: RPL7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK074339.1 | - | 2 | 1790 | ||
AK098059.1 | - | 1 | 1219 |
Gene Symbol: RPS15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023088.1 | - | 5 | 669 |
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209386.1 | - | 2 | 1617 |
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936736.1 | - | 3 | 4074 |
Gene Symbol: SKP1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036619.1 | 1 | 871 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094021.1 | - | 8 | 1440 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 11 | 1529 |
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020677.1 | XP_016876166.1 | 5 | 1631 |
Gene Symbol: TCEANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153035.2 | NP_694580.1 | 5 | 4392 |
Gene Symbol: THNSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092823.1 | - | 1 | 761 |
Gene Symbol: TMEM199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152464.2 | NP_689677.1 | 6 | 1897 | ||
GenBank mRNA | BQ891740.1 | - | 1 | 642 |
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164281.2 | NP_001157753.1 | 9 | 3930 | ||
NM_001164283.2 | NP_001157755.1 | 4 | 2212 | |||
NM_147686.3 | NP_679211.2 | 9 | 3933 | |||
GenBank mRNA | AF136407.1 | - | 9 | 3720 |
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015354.1 | XP_016870843.1 | 6 | 2073 |
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024862.1 | XP_016880351.1 | 5 | 1323 |
Gene Symbol: ZFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310553.1 | - | 6 | 1610 |
Gene Symbol: ZNF891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018666.1 | XP_016874155.1 | 2 | 4253 | ||
XM_017018667.1 | XP_016874156.1 | 2 | 3923 | |||
XM_017018668.1 | XP_016874157.1 | 2 | 4006 |