Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3EAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297590.1 | NP_001284519.1 | 3 | 2917 | ||
NM_012099.1 | NP_036231.1 | 3 | 2911 | |||
XM_017026180.1 | XP_016881669.1 | 3 | 3012 | |||
XM_017026181.1 | XP_016881670.1 | 3 | 3082 | |||
GenBank mRNA | BC038992.1 | - | 3 | 2439 | 6052122 | |
U86751.1 | - | 3 | 2911 |
Gene Symbol: ADIPOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177800.1 | NP_001171271.1 | 4 | 2824 | ||
NM_004797.3 | NP_004788.1 | 3 | 2773 | |||
GenBank mRNA | AL832470.1 | - | 3 | 1025 | ||
AX767959.1 | - | 3 | 2714 | |||
D45371.1 | - | 3 | 2714 |
Gene Symbol: AGFG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012112.1 | XP_016867601.1 | 1 | 227 |
Gene Symbol: AP4B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057567.1 | - | 1 | 991 |
Gene Symbol: BBS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008358.1 | XP_016863847.1 | 17 | 3412 |
Gene Symbol: BRI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011936.1 | XP_016867425.1 | 3 | 3247 |
Gene Symbol: C12orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056213.1 | - | 4 | 2363 |
Gene Symbol: C19orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290149.1 | NP_001277078.1 | 3 | 626 | ||
NM_001290153.1 | NP_001277082.1 | 2 | 304 | |||
NM_001290154.1 | NP_001277083.1 | 2 | 304 | |||
NM_199249.2 | NP_954857.1 | 2 | 304 | |||
GenBank mRNA | BC051842.1 | - | 2 | 348 | 5110472 | |
BG482545.1 | - | 2 | 356 | |||
BG746959.1 | - | 1 | 35 | |||
BG746996.1 | - | 1 | 36 | |||
BG824338.1 | - | 1 | 36 | |||
BM802982.1 | - | 2 | 75 | |||
BQ890364.1 | - | 1 | 33 |
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031910.3 | NP_114116.3 | 3 | 1603 | ||
XM_011529857.1 | XP_011528159.1 | 4 | 4460 | |||
XM_017028569.1 | XP_016884058.1 | 3 | 3752 | |||
XM_017028570.1 | XP_016884059.1 | 5 | 2370 | |||
XM_017028571.1 | XP_016884060.1 | 5 | 4108 | |||
XM_017028572.1 | XP_016884061.1 | 5 | 4097 | |||
XM_017028573.1 | XP_016884062.1 | 5 | 1886 | |||
XM_017028574.1 | XP_016884063.1 | 4 | 1679 | |||
GenBank mRNA | AK226149.1 | - | 2 | 2126 | ||
AY358374.1 | - | 5 | 1740 | |||
BC020551.1 | - | 3 | 1602 | 2900683 | ||
BM722009.1 | - | 1 | 484 | |||
BQ674594.1 | - | 121 |
Gene Symbol: C2orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA653513.1 | - | 1 | 198 |
Gene Symbol: CCDC120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163321.2 | NP_001156793.2 | 11 | 3335 | ||
NM_001163322.2 | NP_001156794.1 | 11 | 3276 | |||
NM_001163323.2 | NP_001156795.1 | 10 | 3765 | |||
NM_001271835.1 | NP_001258764.1 | 10 | 3765 | |||
NM_001271836.1 | NP_001258765.1 | 10 | 4117 | |||
NM_033626.3 | NP_296375.1 | 10 | 3706 | |||
GenBank mRNA | AK056290.1 | - | 1 | 2302 | ||
BC008769.2 | - | 10 | 3687 | 3139519 |
Gene Symbol: CCL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002990.4 | NP_002981.2 | 3 | 1736 | ||
XM_017023531.1 | XP_016879020.1 | 4 | 1756 | |||
GenBank mRNA | BC027952.1 | - | 3 | 1715 | 5220453 | |
U83171.1 | - | 3 | 1726 |
Gene Symbol: CEP104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014704.3 | NP_055519.1 | 22 | 5890 | ||
XM_011542473.2 | XP_011540775.1 | 23 | 6190 | |||
GenBank mRNA | AB011134.1 | - | 23 | 5288 |
Gene Symbol: CFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122686.1 | - | 1 | 595 |
Gene Symbol: CNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540629.2 | XP_011538931.1 | 2 | 5064 | ||
XM_017000261.1 | XP_016855750.1 | 5 | 5756 | |||
GenBank mRNA | EU517121.1 | - | 3 | 4913 |
Gene Symbol: CSRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209321.1 | - | 1 | 2602 |
Gene Symbol: DDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094951.1 | - | 1 | 345 |
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002431.1 | XP_016857920.1 | 1 | 552 |
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255827.3 | XP_005255884.1 | 9 | 3902 | ||
XM_005255829.3 | XP_005255886.1 | 8 | 3539 | |||
XM_017022990.1 | XP_016878479.1 | 9 | 3677 |
Gene Symbol: DNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201366.1 | NP_001188295.1 | 9 | 4354 | ||
NM_031427.3 | NP_113615.2 | 8 | 4308 |
Gene Symbol: EFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311677.1 | - | 3 | 544 |
Gene Symbol: FAM175A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263280.4 | XP_005263337.1 | 10 | 3182 | ||
XM_017008694.1 | XP_016864183.1 | 8 | 2955 |
Gene Symbol: FBXO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648151.1 | - | 5 | 1897 | ||
BX648220.1 | - | 5 | 1894 | |||
BX648644.1 | - | 1 | 1008 |
Gene Symbol: FCHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014311.1 | - | 11 | 1134 |
Gene Symbol: FZD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001466.3 | NP_001457.1 | 1 | 3479 |
Gene Symbol: GABPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323906.1 | NP_001310835.1 | 10 | 1756 | ||
NM_001323911.1 | NP_001310840.1 | 9 | 1642 |
Gene Symbol: GCC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095987.1 | - | 2 | 1251 |
Gene Symbol: GRIK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542786.2 | XP_011541088.1 | 5 | 1165 |
Gene Symbol: GSTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225563.1 | - | 6 | 1677 |
Gene Symbol: IRAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001570.3 | NP_001561.3 | 13 | 2967 | ||
GenBank mRNA | AJ496794.1 | - | 13 | 2908 |
Gene Symbol: KIF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006612.5 | NP_006603.2 | 23 | 4595 | ||
XM_005256424.2 | XP_005256481.1 | 24 | 4586 |
Gene Symbol: KMT2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX640742.1 | - | 1 | 1279 |
Gene Symbol: LETMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123080.1 | - | 1 | 361 |
Gene Symbol: LINC00649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038883.2 | 3 | 2635 | |||
NR_134558.1 | 3 | 2068 |
Gene Symbol: LOC100129724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125769.1 | - | 3 | 1720 |
Gene Symbol: LOC100287792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040021.1 | 4 | 1342 |
Gene Symbol: LOC100507556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129645.1 | - | 1 | 384 |
Gene Symbol: LOC100996251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103777.1 | 3 | 489 | |||
GenBank mRNA | BC037540.1 | - | 3 | 492 |
Gene Symbol: LOC102724392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134268.1 | 3 | 1082 |
Gene Symbol: LOC105374690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043355.1 | - | 7 | 930 |
Gene Symbol: LOC399715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040079.2 | 6 | 2425 | |||
GenBank mRNA | AK128185.1 | - | 7 | 2543 |
Gene Symbol: LOC399886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122071.1 | 1 | 1120 | |||
GenBank mRNA | AK123417.1 | - | 1 | 1121 |
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523206.2 | XP_011521508.1 | 10 | 3586 |
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098831.1 | NP_001092301.1 | 5 | 1354 | ||
NM_178832.3 | NP_849154.1 | 5 | 1443 | |||
XM_011539250.2 | XP_011537552.1 | 4 | 1477 | |||
XM_011539251.2 | XP_011537553.1 | 5 | 1427 | |||
GenBank mRNA | AJ312050.1 | - | 4 | 1348 | ||
AJ431726.1 | - | 5 | 1354 | |||
AK098791.1 | - | 4 | 1245 | |||
AL834132.1 | - | 5 | 1336 | |||
BC010230.2 | - | 5 | 1305 | 3454105 | ||
BC040015.1 | - | 5 | 1306 | 5529065 | ||
DQ338435.1 | - | 5 | 1332 | |||
DQ338436.1 | - | 5 | 1332 |
Gene Symbol: MRPL57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CF890784.1 | - | 1 | 333 |
Gene Symbol: NR4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202234.1 | NP_001189163.1 | 1 | 557 | ||
GenBank mRNA | AK131566.1 | - | 1 | 559 |
Gene Symbol: OR7E5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027688.2 | 2 | 310 | |||
GenBank mRNA | AK055955.1 | - | 2 | 310 |
Gene Symbol: OSBPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647557.1 | - | 1 | 2539 |
Gene Symbol: POLI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007195.2 | NP_009126.2 | 10 | 3658 | ||
GenBank mRNA | AK226057.1 | - | 8 | 3218 |
Gene Symbol: PPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001561.1 | XP_016857050.1 | 4 | 876 | ||
XM_017001564.1 | XP_016857053.1 | 1 | 102 |
Gene Symbol: PPP2R3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037797.1 | - | 1 | 375 |
Gene Symbol: PRKAR2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109996.1 | 2 | 1922 | |||
NR_109997.1 | 3 | 1843 | ||||
GenBank mRNA | AK127190.1 | - | 2 | 1902 |
Gene Symbol: RABGGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127197.1 | - | 1 | 609 |
Gene Symbol: RECK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022897.1 | - | 2 | 634 |
Gene Symbol: RFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052859.3 | NP_443091.1 | 13 | 4637 | ||
GenBank mRNA | AK098811.1 | - | 1 | 1829 |
Gene Symbol: RILPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018959.1 | XP_016874448.1 | 4 | 1393 |
Gene Symbol: RRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028932.1 | - | 4 | 1970 |
Gene Symbol: RUNDC3A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110802.1 | 4 | 3880 | |||
GenBank mRNA | BC051705.1 | - | 3 | 3639 |
Gene Symbol: SIAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017930.1 | XP_016873419.1 | 5 | 1064 |
Gene Symbol: SLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032444.2 | NP_115820.2 | 2 | 374 | ||
XM_011522715.2 | XP_011521017.1 | 2 | 388 | |||
GenBank mRNA | AB075867.1 | - | 1 | 42 | ||
AK122775.1 | - | 2 | 375 | |||
BE887716.1 | - | 322 | ||||
BM541728.1 | - | 2 | 373 |
Gene Symbol: SNHG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003141.3 | 4 | 907 | |||
GenBank mRNA | AJ224167.1 | - | LIT1319 U19HG |
4 | 526 | |
CN337312.1 | - | 2 | 473 | |||
DA203569.1 | - | 1 | 390 |
Gene Symbol: SNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024700.3 | NP_078976.2 | 4 | 3873 |
Gene Symbol: SRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC064351.1 | - | 3 | 868 | 4452218 |
Gene Symbol: ST3GAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124862.1 | - | 2 | 683 |
Gene Symbol: TFAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001288.1 | 1 | 1775 |
Gene Symbol: TLCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164407.1 | NP_001157879.1 | 4 | 4655 |
Gene Symbol: WBSCR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126044.1 | - | 1 | 407 |
Gene Symbol: ZNF652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145365.1 | NP_001138837.1 | 6 | 4949 | ||
NM_014897.2 | NP_055712.1 | 6 | 4804 | |||
XM_005257166.2 | XP_005257223.1 | 5 | 4156 | |||
GenBank mRNA | AB023141.1 | - | 6 | 4803 | ||
BC152440.1 | - | 6 | 4803 | 8862563 |
Gene Symbol: ZNF69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321163.1 | NP_001308092.1 | 5 | 819 | ||
NM_021915.3 | NP_068734.1 | 5 | 777 |
Set Membership: |
G-protein coupled receptor |