Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286703.1 | NP_001273632.1 | 6 | 512 | ||
NM_001286704.1 | NP_001273633.1 | 6 | 690 | |||
NM_001286705.1 | NP_001273634.1 | 7 | 554 | |||
NM_001286706.1 | NP_001273635.1 | 6 | 1188 | |||
NM_016617.3 | NP_057701.1 | 6 | 518 | |||
NR_104584.1 | 7 | 579 | ||||
NR_104585.1 | 5 | 478 | ||||
GenBank mRNA | AF208844.1 | - | 6 | 408 | ||
AK095490.1 | - | 6 | 438 | |||
AL515797.3 | - | 5 | 370 | |||
BC005193.1 | - | 6 | 435 | 3829206 | ||
BG540094.1 | - | 7 | 441 | |||
BI458294.1 | - | 6 | 488 | |||
BI761948.1 | - | 6 | 395 | |||
DA448931.1 | - | 5 | 391 | |||
DA483023.1 | - | 6 | 438 |