Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145638.2 | NP_001139110.2 | 7, 8 | 1200 | ||
NM_015597.5 | NP_056412.5 | 7, 8 | 1200 | |||
XM_011518498.2 | XP_011516800.1 | 7, 8 | 1091 | |||
XM_017014598.1 | XP_016870087.1 | 6, 7 | 912 | |||
GenBank mRNA | AL117478.1 | - | 1, 2 | 40 | ||
AY173053.1 | - | 6, 7 | 911 | |||
BC009979.1 | - | 7, 8 | 1209 | 4121647 | ||
BC017353.1 | - | 7, 8 | 1209 | 5021010 |