Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145638.2 | NP_001139110.2 | 9 | 1385 | ||
NM_015597.5 | NP_056412.5 | 9 | 1385 | |||
XM_011518498.2 | XP_011516800.1 | 9 | 1276 | |||
XM_017014598.1 | XP_016870087.1 | 8 | 1097 | |||
GenBank mRNA | AL117478.1 | - | 3 | 225 | ||
AY173053.1 | - | 8 | 1096 | |||
BC009979.1 | - | 9 | 1394 | 4121647 | ||
BC017353.1 | - | 9 | 1394 | 5021010 |