Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM49B SIRNA Assays ›
Gene Symbol:
FAM49B
Gene Name:
family with sequence similarity 49 member B
Gene Aliases:
BM-009, L1
Chromosome Location:
Chr.8: 129839593 - 130016697 on Build GRCh38
UniGene ID:
Hs.126941
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24737
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM49B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256763.1
NP_001243692.1
7
725
NM_016623.4
NP_057707.3
6
638
NR_046359.1
9
991
NR_046360.1
9
901
NR_046361.1
8
891
XM_005250966.3
XP_005251023.1
5
494
XM_011517108.2
XP_011515410.1
8
787
XM_011517109.2
XP_011515411.1
8
559
XM_011517110.2
XP_011515412.1
7
699
XM_011517111.2
XP_011515413.1
7
517
XM_011517112.2
XP_011515414.1
9
742
XM_011517113.2
XP_011515415.1
8
753
XM_011517114.2
XP_011515416.1
8
659
XM_011517115.2
XP_011515417.1
7
666
XM_011517116.2
XP_011515418.1
8
700
XM_011517117.2
XP_011515419.1
7
669
XM_011517118.2
XP_011515420.1
7
613
XM_011517120.2
XP_011515422.1
6
612
XM_011517121.2
XP_011515423.1
9
624
XM_011517122.2
XP_011515424.1
7
703
XM_011517123.2
XP_011515425.1
7
476
XM_011517124.2
XP_011515426.1
6
591
XM_011517125.2
XP_011515427.1
6
530
XM_017013539.1
XP_016869028.1
9
746
XM_017013540.1
XP_016869029.1
9
605
XM_017013541.1
XP_016869030.1
7
733
XM_017013542.1
XP_016869031.1
9
783
XM_017013543.1
XP_016869032.1
9
883
XM_017013544.1
XP_016869033.1
9
665
XM_017013545.1
XP_016869034.1
10
929
XM_017013546.1
XP_016869035.1
8
696
XM_017013547.1
XP_016869036.1
10
788
XM_017013548.1
XP_016869037.1
8
792
XM_017013549.1
XP_016869038.1
8
655
XM_017013550.1
XP_016869039.1
7
706
XM_017013551.1
XP_016869040.1
9
701
XM_017013552.1
XP_016869041.1
9
746
XM_017013553.1
XP_016869042.1
8
559
XM_017013554.1
XP_016869043.1
10
771
XM_017013555.1
XP_016869044.1
7
726
XM_017013556.1
XP_016869045.1
7
472
XM_017013557.1
XP_016869046.1
8
518
XM_017013558.1
XP_016869047.1
8
663
XM_017013559.1
XP_016869048.1
8
522
XM_017013560.1
XP_016869049.1
6
637
XM_017013561.1
XP_016869050.1
6
389
XM_017013562.1
XP_016869051.1
7
576
GenBank mRNA
AF208851.1
-
8
578
AK001226.1
-
2
144
AK002059.1
-
9
701
AK130372.1
-
6
525
AY364259.1
-
3
193
AY598320.1
-
4
324
BC003599.1
-
7
504
3501828
BC016345.1
-
8
645
4041187
BC017297.2
-
9
873
5017493
BX419336.2
-
7
726
CR749628.1
-
7
471
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0