Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002136.3 | NP_002127.1 | 10 | 1366 | ||
NM_031157.3 | NP_112420.1 | 11 | 1522 | |||
XM_017019251.1 | XP_016874740.1 | 9 | 1285 | |||
GenBank mRNA | AK129814.1 | - | 10 | 1308 | ||
AK298176.1 | - | 7 | 865 | |||
BC002355.2 | - | 10 | 1287 | 2821751 | ||
BC004945.1 | - | 1 | 1184 | |||
BC009600.1 | - | 10 | 1309 | 4134638 | ||
BC012158.1 | - | 10 | 1269 | 4564655 | ||
BC020442.2 | - | 1 | 1276 | |||
BC033714.1 | - | 10 | 1298 | 4420039 | ||
BC035253.1 | - | 6 | 725 | |||
BC052296.1 | - | 10 | 1278 | 6297642 | ||
BC071945.1 | - | 1 | 1298 | 5475712 | ||
BC073162.1 | - | 1 | 1292 | 6154972 | ||
BC074502.1 | - | 10 | 1291 | 3455742 | ||
BC103707.1 | - | 9 | 1117 | 6154800 | ||
BU501222.1 | - | 1 | 739 | |||
U00947.1 | - | 10 | 1255 | |||
X04347.1 | - | 7 | 875 | |||
X06747.1 | - | 10 | 1332 |
Gene Symbol: HNRNPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011724.1 | NP_001011724.1 | 7 | 2320 | ||
NM_001011725.1 | NP_001011725.1 | 6 | 2179 | |||
GenBank mRNA | AK091082.1 | - | 1 | 2032 | ||
AK094423.1 | - | 6 | 2070 | |||
AK123159.1 | - | 5 | 1859 | |||
AK126454.1 | - | 1 | 2954 | |||
BC082242.1 | - | 6 | 2110 | |||
BC108266.1 | - | 1 | 1308 | 4778431 |
Gene Symbol: HNRNPA1P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002944.2 | 1 | 1304 |
Gene Symbol: SMARCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003601.3 | NP_003592.3 | 24 | 5961 | ||
GenBank mRNA | AK123439.1 | - | 1 | 32 |