Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002136.3 | NP_002127.1 | 4 | 429 | ||
NM_031157.3 | NP_112420.1 | 4 | 429 | |||
NR_135167.1 | 4 | 429 | ||||
XM_005268826.1 | XP_005268883.1 | 4 | 423 | |||
XM_017019251.1 | XP_016874740.1 | 4 | 423 | |||
GenBank mRNA | AK129814.1 | - | 4 | 371 | ||
AK291113.1 | - | 4 | 369 | |||
AK298176.1 | - | 4 | 372 | |||
AK308679.1 | - | 3 | 323 | |||
BC002355.2 | - | 4 | 350 | 2821751 | ||
BC004945.1 | - | 1 | 246 | |||
BC009600.1 | - | 4 | 372 | 4134638 | ||
BC012158.1 | - | 4 | 332 | 4564655 | ||
BC020442.2 | - | 1 | 339 | |||
BC033714.1 | - | 4 | 361 | 4420039 | ||
BC052296.1 | - | 4 | 341 | 6297642 | ||
BC070315.1 | - | 4 | 371 | 4101267 | ||
BC071945.1 | - | 1 | 361 | 5475712 | ||
BC073162.1 | - | 1 | 355 | 6154972 | ||
BC074502.1 | - | 4 | 354 | 3455742 | ||
BC103707.1 | - | 4 | 339 | 6154800 | ||
BC121133.2 | - | 4 | 355 | 40120894 | ||
BQ277208.1 | - | 429 | ||||
CD705336.1 | - | 4 | 446 | |||
DC358304.1 | - | 4 | 525 | |||
U00947.1 | - | 4 | 319 | |||
X06747.1 | - | 4 | 396 | |||
X79536.1 | - | 4 | 337 |
Gene Symbol: HNRNPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011724.1 | NP_001011724.1 | 7 | 1384 | ||
NM_001011725.1 | NP_001011725.1 | 6 | 1243 | |||
GenBank mRNA | AK091082.1 | - | 1 | 1096 | ||
AK094423.1 | - | 6 | 1134 | |||
AK123159.1 | - | 5 | 923 | |||
AK126454.1 | - | 1 | 2018 | |||
BC082242.1 | - | 6 | 1174 | |||
BC108266.1 | - | 1 | 372 | 4778431 |
Gene Symbol: HNRNPA1P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002944.2 | 1 | 370 |
Gene Symbol: XKRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530954.2 | XP_011529256.1 | 4 | 2792 |