Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 6336 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 5402 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 6203 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 5890 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 6465 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 6729 | |||
NM_003879.5 | NP_003870.4 | 10 | 6308 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 7203 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 6172 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 6251 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 7138 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 9155 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 7011 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 6349 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 5974 |
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 63549 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 1683 | |||
GenBank mRNA | BC036758.2 | - | 7 | 1695 | 5563071 |
Gene Symbol: SMC5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936772.1 | - | 3 | 5345 |
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648230.1 | - | 7 | 2704 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 1561 | |||
GenBank mRNA | AK056558.1 | - | 5 | 1381 |