Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172241.2 | NP_758441.2 | 1 | 1058 | ||
GenBank mRNA | AF273058.1 | - | 1 | 1066 | ||
BC031065.1 | - | 1 | 1060 | |||
BC132871.1 | - | 1 | 991 | 40146893 |
Gene Symbol: CTAGE11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027466.1 | 1 | 1076 | |||
GenBank mRNA | AL133018.1 | - | 1 | 1076 |
Gene Symbol: CTAGE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008747.2 | NP_001008747.1 | 1 | 1067 | ||
GenBank mRNA | BC146818.1 | - | 1 | 1067 | 8860423 |
Gene Symbol: CTAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198495.2 | NP_940897.2 | 1 | 1099 | ||
GenBank mRNA | AF338232.1 | - | 1 | 428 |
Gene Symbol: CTAGE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178561.4 | NP_848656.2 | 1 | 1142 | ||
GenBank mRNA | BC101323.1 | - | 1 | 400 |
Gene Symbol: CTAGE7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_044994.1 | 1 | 1075 | |||
GenBank mRNA | BC036527.1 | - | 1 | 1078 |
Gene Symbol: CTAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278507.1 | NP_001265436.1 | 1 | 1584 | ||
GenBank mRNA | AK292236.1 | - | 1 | 1088 |
Gene Symbol: CTAGE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145659.1 | NP_001139131.1 | 1 | 1050 | ||
GenBank mRNA | BC101322.1 | - | 1 | 400 |