Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173462.3 | NP_775733.3 | 26 | 4649 | ||
XM_011537290.2 | XP_011535592.1 | 27 | 4730 | |||
XM_011537291.2 | XP_011535593.1 | 28 | 4777 | |||
XM_011537292.2 | XP_011535594.1 | 27 | 4653 | |||
XM_011537293.2 | XP_011535595.1 | 26 | 4696 | |||
XM_011537294.2 | XP_011535596.1 | 26 | 4654 | |||
XM_011537295.2 | XP_011535597.1 | 24 | 4412 | |||
XM_017021749.1 | XP_016877238.1 | 24 | 4459 | |||
XM_017021750.1 | XP_016877239.1 | 26 | 4652 | |||
GenBank mRNA | AK021713.1 | - | 1 | 470 | ||
AK125658.1 | - | 6 | 1829 | |||
AK127354.1 | - | 12 | 2798 | |||
AK131073.1 | - | 19 | 3943 | |||
AL110280.1 | - | 8 | 3500 | |||
AY358330.1 | - | 10 | 2554 | |||
BC042057.1 | - | 7 | 635 | |||
BX537757.1 | - | 12 | 2774 |
Gene Symbol: ACBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042473.3 | NP_001035938.1 | 13 | 3019 | ||
NM_001271512.3 | NP_001258441.1 | 12 | 3333 | |||
NM_001301251.1 | NP_001288180.1 | 12 | 2898 | |||
NM_001301252.1 | NP_001288181.1 | 11 | 3212 | |||
NM_001301253.1 | NP_001288182.1 | 12 | 2888 | |||
NM_001301254.1 | NP_001288183.1 | 10 | 3008 | |||
NM_145698.4 | NP_663736.2 | 13 | 3144 | |||
XM_006717528.3 | XP_006717591.3 | 14 | 9577 | |||
XM_006717530.3 | XP_006717593.1 | 14 | 3176 | |||
XM_006717531.2 | XP_006717594.1 | 14 | 3186 | |||
XM_006717535.2 | XP_006717598.1 | 14 | 3043 | |||
XM_017016886.1 | XP_016872375.1 | 13 | 9544 | |||
XM_017016892.1 | XP_016872381.1 | 13 | 3060 | |||
XM_017016894.1 | XP_016872383.1 | 12 | 9340 | |||
XM_017016897.1 | XP_016872386.1 | 13 | 2968 | |||
XM_017016899.1 | XP_016872388.1 | 12 | 2935 | |||
XM_017016902.1 | XP_016872391.1 | 13 | 2960 | |||
XM_017016903.1 | XP_016872392.1 | 13 | 2945 | |||
XM_017016906.1 | XP_016872395.1 | 12 | 2807 | |||
XM_017016907.1 | XP_016872396.1 | 12 | 2940 | |||
XM_017016908.1 | XP_016872397.1 | 12 | 2929 | |||
GenBank mRNA | AB082527.1 | - | 12 | 2847 | ||
AL133064.1 | - | 7 | 2225 |
Gene Symbol: ACTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296991.1 | - | 4 | 1069 |
Gene Symbol: ACVRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000020.2 | NP_000011.2 | 10 | 2799 | ||
NM_001077401.1 | NP_001070869.1 | 9 | 2662 | |||
XM_005269235.2 | XP_005269292.1 | 11 | 2801 | |||
XM_011539008.1 | XP_011537310.1 | 7 | 2345 | |||
GenBank mRNA | AK056725.1 | - | 1 | 497 |
Gene Symbol: ALDH6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278593.1 | NP_001265522.1 | 12 | 3373 | ||
NM_001278594.1 | NP_001265523.1 | 12 | 3575 | |||
NM_005589.3 | NP_005580.1 | 12 | 3412 | |||
XM_017021331.1 | XP_016876820.1 | 10 | 3263 |
Gene Symbol: ASCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF132952.1 | - | 1 | 144 | ||
AY013290.1 | - | 1 | 144 |
Gene Symbol: COX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320975.1 | NP_001307904.1 | 8 | 2460 | ||
NM_001320976.1 | NP_001307905.1 | 9 | 2532 | |||
NM_078470.5 | NP_510870.1 | 9 | 2615 | |||
GenBank mRNA | BC002382.2 | - | 9 | 2014 | 2961532 | |
BC013403.2 | - | 9 | 2014 | 2961532 | ||
BC078161.1 | - | 9 | 2027 | 6150426 | ||
BX537557.1 | - | 9 | 2614 |
Gene Symbol: CROCCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054737.1 | - | 1 | 301 | ||
AK090414.1 | - | 11 | 3936 |
Gene Symbol: CSAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126934.1 | - | 1 | 3045 |
Gene Symbol: DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164539.1 | NP_001158011.1 | 10 | 2177 | ||
GenBank mRNA | FJ804196.1 | - | 10 | 2177 | ||
FJ804197.1 | - | 9 | 2107 | |||
FJ804198.1 | - | 9 | 2026 |
Gene Symbol: FAM118A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028855.1 | XP_016884344.1 | 6 | 1251 |
Gene Symbol: FAM186A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC140703.1 | - | 5 | 1901 |
Gene Symbol: FBXO22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521447.2 | XP_011519749.1 | 8 | 4954 |
Gene Symbol: HS3ST3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006041.2 | NP_006032.1 | 2 | 2412 | ||
GenBank mRNA | BC063301.1 | - | 2 | 2322 | 30343352 |
Gene Symbol: LOC101928514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055500.1 | - | 2 | 2116 |
Gene Symbol: MOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003090.2 | 6 | 3268 | |||
NR_103504.1 | 7 | 3610 | ||||
NR_103505.1 | 7 | 3648 | ||||
GenBank mRNA | AK095118.1 | - | 1 | 341 | ||
BC035141.1 | - | 1 | 34 |
Gene Symbol: NFATC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057574.1 | - | 1 | 406 |
Gene Symbol: NFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091049.1 | - | 1 | 49 |
Gene Symbol: NICN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032316.3 | NP_115692.1 | 6 | 1951 |
Gene Symbol: NSUN5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094312.1 | - | 1 | 469 | ||
AK126486.1 | - | 3 | 2443 | |||
AK128374.1 | - | 1 | 681 |
Gene Symbol: NUDT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301022.1 | NP_001287951.1 | 5 | 2083 | ||
NM_001301023.1 | NP_001287952.1 | 5 | 2080 | |||
NM_001301024.1 | NP_001287953.1 | 5 | 1983 | |||
NM_019094.5 | NP_061967.3 | 5 | 2338 | |||
NM_199040.3 | NP_950241.1 | 5 | 2341 | |||
XM_011537794.2 | XP_011536096.1 | 5 | 1881 | |||
XM_011537795.2 | XP_011536097.1 | 6 | 3893 | |||
XM_011537796.2 | XP_011536098.1 | 6 | 2022 | |||
GenBank mRNA | AB007956.2 | - | 6 | 2137 | ||
AF191651.1 | - | 5 | 1931 | |||
AF191652.2 | - | 5 | 1929 | |||
AF191654.2 | - | 1 | 17 | |||
BC012069.1 | - | 5 | 2130 | 4650281 | ||
CR749445.1 | - | 5 | 2053 |
Gene Symbol: NUDT4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002212.4 | 1 | 2295 | |||
GenBank mRNA | AF311215.1 | - | LIT2089 NUDT4 pseudogene |
5 | 2217 |
Gene Symbol: NUDT4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104005.1 | 1 | 2339 | |||
GenBank mRNA | AK074343.1 | - | 5 | 2341 |
Gene Symbol: PKD1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832135.1 | - | 1 | 2549 |
Gene Symbol: PLBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011538023.2 | XP_011536325.1 | 11 | 2833 | ||
XM_017018977.1 | XP_016874466.1 | 11 | 2946 |
Gene Symbol: POLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936627.1 | - | 14 | 3052 |
Gene Symbol: RAB36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004914.3 | NP_004905.2 | 11 | 2554 | ||
XM_005261859.4 | XP_005261916.1 | 12 | 2638 | |||
XM_006724381.3 | XP_006724444.1 | 12 | 2621 | |||
XM_006724382.3 | XP_006724445.1 | 11 | 2546 | |||
XM_011530544.2 | XP_011528846.1 | 12 | 2622 | |||
XM_011530547.2 | XP_011528849.1 | 10 | 2468 | |||
XM_011530552.2 | XP_011528854.1 | 8 | 2577 | |||
XM_017029104.1 | XP_016884593.1 | 11 | 2542 | |||
XM_017029105.1 | XP_016884594.1 | 10 | 2484 | |||
XM_017029106.1 | XP_016884595.1 | 9 | 2281 | |||
XM_017029107.1 | XP_016884596.1 | 10 | 2317 | |||
GenBank mRNA | AB023061.1 | - | 11 | 2554 | ||
AF133588.1 | - | 11 | 2517 | |||
AK296052.1 | - | 5 | 772 |
Gene Symbol: SH2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291424.1 | NP_001278353.1 | 7 | 2948 | ||
NM_005475.2 | NP_005466.1 | 8 | 3871 | |||
XM_005253818.4 | XP_005253875.1 | 9 | 4015 | |||
XM_005253819.4 | XP_005253876.1 | 8 | 3898 | |||
XM_006719180.3 | XP_006719243.1 | 7 | 3500 | |||
XM_011537719.2 | XP_011536021.1 | 9 | 4018 | |||
XM_011537720.2 | XP_011536022.1 | 9 | 3683 | |||
XM_011537721.2 | XP_011536023.1 | 7 | 5483 | |||
GenBank mRNA | AB208911.1 | - | 7 | 3108 | ||
AF055581.1 | - | 8 | 3869 | |||
AK025666.1 | - | 1 | 878 | |||
AK025745.1 | - | 1 | 1715 | |||
BC015786.1 | - | 1 | 208 |
Gene Symbol: SLC25A53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012755.4 | NP_001012773.2 | 2 | 2449 | ||
XM_005262129.4 | XP_005262186.1 | 2 | 2395 | |||
XM_011530952.2 | XP_011529254.1 | 3 | 2592 | |||
XM_011530953.2 | XP_011529255.1 | 3 | 2423 | |||
XM_017029514.1 | XP_016885003.1 | 3 | 3234 | |||
XM_017029515.1 | XP_016885004.1 | 4 | 2742 | |||
XM_017029516.1 | XP_016885005.1 | 3 | 3365 | |||
GenBank mRNA | AL833609.1 | - | 2 | 2450 | ||
BC140812.1 | - | 1 | 2308 | 9021680 | ||
BC140814.1 | - | 1 | 2309 | 9021682 |
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI711397.1 | - | 1 | 202 |
Gene Symbol: TEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091669.1 | - | 1 | 1062 |
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021156.3 | NP_066979.2 | 8 | 3925 | ||
GenBank mRNA | AB032988.1 | - | 1 | 3009 | ||
BC044777.1 | - | 8 | 3924 |
Gene Symbol: ZNF528-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537909.1 | - | 4 | 4244 |