Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032121.5 | NP_115497.4 | 4 | 578 | ||
GenBank mRNA | AK027632.1 | - | 2 | 95 | ||
AK075394.1 | - | 4 | 441 | |||
AK223131.1 | - | 4 | 441 | |||
AK294943.1 | - | 4 | 603 | |||
AK296743.1 | - | 4 | 441 | |||
AK314309.1 | - | 4 | 441 | |||
AL136636.1 | - | 4 | 449 | |||
AY358691.1 | - | 4 | 428 | |||
BC041014.1 | - | 4 | 425 | 3834858 | ||
BC060842.1 | - | 4 | 441 | 30338916 | ||
DQ000004.1 | - | 4 | 449 |