Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145666.1 | NP_001139138.1 | 26 | 6975 | ||
NM_001145667.1 | NP_001139139.1 | 26 | 8047 | |||
NM_012201.5 | NP_036333.2 | 27 | 7008 | |||
NR_027264.1 | 27 | 8156 | ||||
NR_027265.1 | 21 | 7044 | ||||
GenBank mRNA | AK057359.1 | - | 1 | 650 | ||
BX648396.1 | - | 1 | 1183 |
Gene Symbol: AGTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251847.3 | XP_005251904.1 | 26 | 5293 |
Gene Symbol: ARFGEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144626.1 | - | 24 | 4075 |
Gene Symbol: ASPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024661.1 | XP_016880150.1 | 8 | 2723 |
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627020.1 | - | 1 | 773 |
Gene Symbol: BTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016873.1 | XP_016872362.1 | 1 | 32 |
Gene Symbol: C1orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095622.1 | - | 1 | 1065 |
Gene Symbol: CA5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057568.1 | - | 1 | 1678 |
Gene Symbol: CBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127321.1 | NP_001120793.1 | 5 | 6879 | ||
NM_001127322.1 | NP_001120794.1 | 5 | 7077 | |||
NM_012117.2 | NP_036249.1 | 5 | 6925 | |||
GenBank mRNA | AK056315.1 | - | 1 | 1060 |
Gene Symbol: CD3EAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297590.1 | NP_001284519.1 | 3 | 2477 | ||
NM_012099.1 | NP_036231.1 | 3 | 2471 | |||
XM_017026180.1 | XP_016881669.1 | 3 | 2572 | |||
XM_017026181.1 | XP_016881670.1 | 3 | 2642 | |||
GenBank mRNA | BC038992.1 | - | 3 | 1999 | 6052122 | |
U86751.1 | - | 3 | 2471 |
Gene Symbol: CDCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022842.4 | NP_073753.3 | 9 | 4417 | ||
XM_011534024.1 | XP_011532326.1 | 9 | 4375 | |||
XM_017007070.1 | XP_016862559.1 | 8 | 4315 | |||
GenBank mRNA | AF468010.1 | - | 9 | 4392 | ||
AK023834.1 | - | 7 | 3995 | |||
AY026461.1 | - | 9 | 4363 | |||
AY167484.1 | - | 9 | 4374 |
Gene Symbol: CLSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190481.1 | NP_001177410.1 | 24 | 4738 | ||
NM_022111.3 | NP_071394.2 | 25 | 4930 | |||
XM_011541938.2 | XP_011540240.1 | 25 | 4953 | |||
XM_011541941.2 | XP_011540243.1 | 24 | 4785 |
Gene Symbol: CPT1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB208884.1 | - | 9 | 1760 |
Gene Symbol: CREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510777.2 | XP_011509079.1 | 3 | 1428 | ||
XM_017003565.1 | XP_016859054.1 | 3 | 1821 |
Gene Symbol: CRKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005207.3 | NP_005198.1 | 3 | 2049 | ||
GenBank mRNA | BC043500.1 | - | 3 | 1990 | 6046673 |
Gene Symbol: DAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR933698.1 | - | 6 | 1566 |
Gene Symbol: DDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127128.1 | - | 1 | 1966 |
Gene Symbol: DENND2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX649075.1 | - | 15 | 1822 |
Gene Symbol: DTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080664.2 | NP_542395.1 | 3 | 938 | ||
GenBank mRNA | AK022703.1 | - | 1 | 316 | ||
AK055367.1 | - | 3 | 925 | |||
BC010618.1 | - | 3 | 889 | 3870759 |
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC035000.1 | - | 4 | 1473 | 5269106 |
Gene Symbol: ENOSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309035.1 | - | 8 | 1048 |
Gene Symbol: ERCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005236.2 | NP_005227.1 | 11 | 4434 | ||
XM_011522424.2 | XP_011520726.1 | 12 | 4591 | |||
XM_011522427.1 | XP_011520729.1 | 6 | 3571 |
Gene Symbol: FCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318508.1 | NP_001305437.1 | 8 | 1715 | ||
NM_015962.4 | NP_057046.1 | 8 | 1679 | |||
XM_005267730.3 | XP_005267787.1 | 8 | 1811 | |||
XM_011536815.2 | XP_011535117.1 | 7 | 1636 | |||
XM_011536816.2 | XP_011535118.1 | 7 | 1636 | |||
GenBank mRNA | BX647810.1 | - | 8 | 1705 |
Gene Symbol: GCSHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092810.1 | - | 1 | 377 |
Gene Symbol: HTATSF1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128391.1 | - | 3 | 1067 | ||
BX648511.1 | - | 2 | 1208 |
Gene Symbol: JPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056172.1 | - | 5 | 671 |
Gene Symbol: LINC00189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027072.2 | 1 | 304 |
Gene Symbol: LINC01012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038292.1 | 3 | 1744 | |||
GenBank mRNA | BC035101.1 | - | 3 | 1745 |
Gene Symbol: LOC105371932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025492.1 | XP_016880981.1 | 4 | 1562 |
Gene Symbol: LOC105374013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513341.2 | XP_011511643.1 | 1 | 162 |
Gene Symbol: LOC401490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052949.1 | - | 3 | 3459 |
Gene Symbol: LPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167671.2 | NP_001161143.1 | 11 | 10347 | ||
NM_001167672.2 | NP_001161144.1 | 10 | 9875 | |||
NM_005578.4 | NP_005569.1 | 11 | 10404 | |||
XM_005247446.4 | XP_005247503.1 | 11 | 10381 | |||
XM_005247451.4 | XP_005247508.1 | 11 | 10381 | |||
XM_011512820.2 | XP_011511122.1 | 13 | 10648 | |||
XM_011512833.2 | XP_011511135.1 | 12 | 10438 | |||
XM_011512834.2 | XP_011511136.1 | 12 | 10529 | |||
XM_017006377.1 | XP_016861866.1 | 13 | 10510 | |||
XM_017006378.1 | XP_016861867.1 | 12 | 10526 | |||
XM_017006379.1 | XP_016861868.1 | 12 | 10661 |
Gene Symbol: LRCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128189.1 | - | 1 | 1313 |
Gene Symbol: LYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000239.2 | NP_000230.1 | 4 | 831 | ||
GenBank mRNA | AK130127.1 | - | 1 | 198 | ||
AK130149.1 | - | 4 | 836 | |||
BC004147.2 | - | 4 | 795 | 2959387 | ||
J03801.1 | - | 4 | 802 | |||
M19045.1 | - | 4 | 798 |
Gene Symbol: MAPK1IP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537362.2 | XP_011535664.2 | 1 | 62 |
Gene Symbol: MCF2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647790.1 | - | 3 | 7035 |
Gene Symbol: MPST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB208824.1 | - | 3 | 3072 |
Gene Symbol: MTPAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018109.3 | NP_060579.3 | 9 | 2893 |
Gene Symbol: OGFOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024648.2 | NP_078924.1 | 9 | 3531 | ||
NM_175902.4 | NP_787098.3 | 10 | 3744 | |||
NR_033265.1 | 10 | 3736 |
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU361469.1 | - | 1 | 58 | ||
KC357595.1 | - | 2 | 58 |
Gene Symbol: PCBP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033872.1 | 4 | 304 | |||
GenBank mRNA | BC040598.1 | - | 4 | 304 |
Gene Symbol: PDLIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256429.1 | NP_001243358.1 | 3 | 292 | ||
NR_046186.1 | 3 | 717 | ||||
GenBank mRNA | AY345240.1 | - | 3 | 264 |
Gene Symbol: PIGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002644.3 | NP_002635.2 | 11 | 3292 | ||
XM_011509629.1 | XP_011507931.1 | 11 | 3203 | |||
GenBank mRNA | AF272149.2 | - | 11 | 3288 | ||
AK310199.1 | - | 1 | 741 | |||
AY178835.1 | - | 1 | 817 | |||
CR749533.1 | - | 12 | 3365 |
Gene Symbol: PLA2G15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522979.2 | XP_011521281.1 | 3 | 355 | ||
XM_011522980.2 | XP_011521282.2 | 3 | 351 |
Gene Symbol: PLA2G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648966.1 | - | 1 | 3878 |
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA437806.1 | - | 1 | 526 |
Gene Symbol: RDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526408.2 | XP_011524710.1 | 7 | 2614 |
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW080436.1 | - | 1 | 205 |
Gene Symbol: RPAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098491.1 | - | 1 | 256 |
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093144.1 | - | 1 | 1490 |
Gene Symbol: SEMA4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126512.1 | - | 1 | 509 |
Gene Symbol: SGSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209900.1 | - | 5 | 1691 |
Gene Symbol: SHISA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL110201.1 | - | 1 | 2208 |
Gene Symbol: SLC16A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201546.1 | NP_001188475.1 | 8 | 1752 | ||
NM_001201547.1 | NP_001188476.1 | 8 | 1763 | |||
NM_001201548.1 | NP_001188477.1 | 7 | 1639 | |||
NM_001201549.1 | NP_001188478.1 | 8 | 1392 | |||
NM_001319220.1 | NP_001306149.1 | 7 | 1259 | |||
NM_004696.2 | NP_004687.1 | 9 | 1896 | |||
XM_005271317.4 | XP_005271374.1 | 10 | 2914 | |||
XM_006711033.3 | XP_006711096.1 | 7 | 2505 | |||
GenBank mRNA | AK129985.1 | - | 9 | 1838 | ||
AK290720.1 | - | 8 | 1697 | |||
AL833619.1 | - | 8 | 1692 | |||
BC021664.1 | - | 8 | 1361 | 4050756 | ||
BX647948.1 | - | 7 | 1223 | |||
U59185.1 | - | 9 | 1828 |
Gene Symbol: SRP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005265765.4 | XP_005265822.3 | 17 | 2915 |
Gene Symbol: SRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095112.1 | - | 1 | 131 | ||
AK126318.1 | - | 1 | 1502 |
Gene Symbol: STYXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308828.1 | - | 8, 9 | 780 |
Gene Symbol: TGFBRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142621.2 | NP_001136093.1 | 12 | 4643 | ||
NM_004257.5 | NP_004248.2 | 12 | 4646 | |||
XM_017005339.1 | XP_016860828.1 | 11 | 4139 | |||
GenBank mRNA | BC019841.1 | - | 1 | 807 |
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133920.1 | 4 | 481 | |||
XM_005262774.3 | XP_005262831.1 | 6 | 2585 | |||
GenBank mRNA | AK056959.1 | - | 4 | 441 | ||
AK308914.1 | - | 6 | 602 | |||
DA995979.1 | - | 4 | 471 |
Gene Symbol: TMEM184C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018241.2 | NP_060711.2 | 10 | 2238 | ||
GenBank mRNA | AF305823.1 | - | 3 | 913 | ||
AK000169.1 | - | 7 | 1390 | |||
AK001708.1 | - | 10 | 2180 | |||
BC008120.1 | - | 2 | 769 |
Gene Symbol: TMIGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU629751.1 | - | 1 | 190 | ||
HY260382.1 | - | 1 | 204 |
Gene Symbol: TNRC6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721996.3 | XP_006722059.3 | 1 | 2972 | ||
XM_006721997.3 | XP_006722060.3 | 1 | 2972 | |||
XM_006722000.3 | XP_006722063.3 | 1 | 2972 | |||
XM_017024906.1 | XP_016880395.1 | 1 | 2972 | |||
XM_017024907.1 | XP_016880396.1 | 1 | 2972 | |||
XM_017024908.1 | XP_016880397.1 | 1 | 2972 |
Gene Symbol: TRAF3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139490.1 | NP_001132962.1 | 15 | 3233 | ||
NM_015650.3 | NP_056465.2 | 17 | 3431 | |||
XM_006712414.2 | XP_006712477.1 | 15 | 3285 | |||
XM_011510944.2 | XP_011509246.1 | 18 | 3588 | |||
XM_011510945.2 | XP_011509247.1 | 18 | 3549 | |||
XM_011510946.2 | XP_011509248.1 | 17 | 3516 | |||
XM_011510947.2 | XP_011509249.1 | 18 | 3575 | |||
XM_011510948.2 | XP_011509250.1 | 16 | 3390 | |||
XM_011510950.2 | XP_011509252.1 | 10 | 2391 | |||
XM_017003789.1 | XP_016859278.1 | 18 | 3585 | |||
GenBank mRNA | BC059174.1 | - | 15 | 3236 | 6191726 |
Gene Symbol: TRMT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001504.1 | XP_016856993.1 | 10 | 862 | ||
XM_017001505.1 | XP_016856994.1 | 10 | 851 |
Gene Symbol: USP6NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125732.1 | 3 | 473 | |||
GenBank mRNA | AK091487.1 | - | 2 | 78 | ||
AK311094.1 | - | 2 | 121 | |||
AK311453.1 | - | 3 | 268 | |||
BC010351.1 | - | 3 | 351 |
Gene Symbol: WDR74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274055.2 | XP_005274112.1 | 1 | 1369 |
Gene Symbol: ZFYVE19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209224.1 | - | 793 |
Gene Symbol: ZMAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011657.3 | NP_001011657.2 | 2 | 388 |
Gene Symbol: ZNF383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258585.2 | XP_005258642.1 | 6 | 2423 | ||
XM_005258587.2 | XP_005258644.1 | 5 | 2300 | |||
XM_005258588.2 | XP_005258645.1 | 6 | 2062 | |||
XM_011526586.2 | XP_011524888.1 | 7 | 3168 | |||
XM_011526587.2 | XP_011524889.1 | 6 | 3135 | |||
XM_011526588.2 | XP_011524890.1 | 5 | 3317 | |||
XM_011526589.2 | XP_011524891.1 | 5 | 1998 | |||
XM_011526590.2 | XP_011524892.1 | 4 | 1963 | |||
XM_017026422.1 | XP_016881911.1 | 7 | 2574 | |||
XM_017026423.1 | XP_016881912.1 | 7 | 3168 | |||
XM_017026424.1 | XP_016881913.1 | 5 | 2214 |
Gene Symbol: ZSCAN22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526917.2 | XP_011525219.1 | 1 | 545 |