Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD99L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184808.1 | NP_001171737.1 | 1, 2 | 289 | ||
NM_001242614.1 | NP_001229543.1 | 1, 2 | 289 | |||
NM_031462.3 | NP_113650.2 | 1, 2 | 289 | |||
NM_134445.3 | NP_604394.1 | 1, 2 | 289 | |||
NM_134446.3 | NP_604395.1 | 1, 2 | 289 | |||
XM_011531202.2 | XP_011529504.1 | 1, 2 | 190 | |||
XM_017029890.1 | XP_016885379.1 | 1, 2 | 153 | |||
XM_017029891.1 | XP_016885380.1 | 1, 2 | 287 | |||
GenBank mRNA | AK125020.1 | - | 1, 2 | 100 | ||
AK290200.1 | - | 1, 2 | 65 | |||
AK291375.1 | - | 1, 2 | 102 | |||
AK293244.1 | - | 1, 2 | 95 | |||
AK295217.1 | - | 1, 2 | 107 | |||
AL136580.1 | - | 1, 2 | 179 | |||
AY078165.1 | - | 1, 2 | 97 | |||
AY078166.1 | - | 1, 2 | 97 | |||
AY078167.1 | - | 1, 2 | 97 | |||
AY358837.1 | - | 1, 2 | 162 | |||
BC025729.1 | - | 1, 2 | 103 | 5203087 | ||
BC030536.1 | - | 1, 2 | 149 | 5242536 | ||
DC323066.1 | - | 1, 2 | 289 | |||
KU178797.1 | - | 1, 2 | 61 |