Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145463.1 | NP_001138935.1 | 8 | 867 | ||
XM_017027449.1 | XP_016882938.1 | 8 | 877 | |||
XM_017027450.1 | XP_016882939.1 | 8 | 875 | |||
XM_017027451.1 | XP_016882940.1 | 8 | 873 | |||
GenBank mRNA | AK304479.1 | - | 3 | 1727 | ||
AL833382.1 | - | 8 | 824 | |||
BC014974.1 | - | 8 | 821 | 4865479 |
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833238.1 | - | 3 | 2371 |
Gene Symbol: ARHGEF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX404969.2 | - | 1 | 231 |
Gene Symbol: ARMC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511916.2 | XP_011510218.1 | 20 | 7144 |
Gene Symbol: BUD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130600.1 | - | 1 | 108 |
Gene Symbol: C11orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094203.1 | - | 4 | 1198 |
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243077.1 | NP_001230006.1 | 4 | 1443 | ||
NM_001243078.1 | NP_001230007.1 | 3 | 1377 | |||
NM_006139.3 | NP_006130.1 | 4 | 1734 | |||
XM_011512194.2 | XP_011510496.1 | 4 | 1569 | |||
XM_011512195.2 | XP_011510497.1 | 5 | 1311 | |||
XM_011512197.2 | XP_011510499.1 | 5 | 1333 | |||
GenBank mRNA | DA939711.1 | - | 1 | 183 |
Gene Symbol: CRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040647.1 | NP_001035737.1 | 5 | 1319 | ||
NM_001040648.1 | NP_001035738.1 | 3 | 1187 | |||
NM_001142414.1 | NP_001135886.1 | 5 | 1263 | |||
NM_014478.4 | NP_055293.1 | 6 | 1418 | |||
NR_024548.1 | 7 | 1540 | ||||
GenBank mRNA | AK023295.1 | - | 1 | 513 | ||
BC040107.1 | - | 5 | 1265 | 5585294 | ||
BC052570.1 | - | 7 | 1754 | |||
BX647867.1 | - | 5 | 1258 |
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001424.5 | NP_001415.1 | 5 | 2035 | ||
XM_006720864.3 | XP_006720927.1 | 5 | 2029 | |||
GenBank mRNA | AF190163.1 | - | 1 | 454 | ||
DA646653.1 | - | 1 | 134 |
Gene Symbol: GPATCH2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB032978.1 | - | 6 | 6156 |
Gene Symbol: HOMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720775.3 | XP_006720838.1 | 10 | 8139 |
Gene Symbol: IL17RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289905.1 | NP_001276834.1 | 12 | 3386 | ||
NM_014339.6 | NP_055154.3 | 13 | 3488 |
Gene Symbol: IL23R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144701.2 | NP_653302.2 | 11 | 2556 | ||
GenBank mRNA | AF461422.1 | - | 11 | 2556 |
Gene Symbol: IVD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159508.1 | NP_001152980.1 | 11 | 2140 | ||
NM_002225.3 | NP_002216.2 | 12 | 2230 | |||
XM_017022152.1 | XP_016877641.1 | 12 | 2011 | |||
XM_017022154.1 | XP_016877643.1 | 12 | 2152 | |||
XM_017022156.1 | XP_016877645.1 | 12 | 2297 | |||
GenBank mRNA | AF070531.1 | - | 4 | 1264 | ||
AK022777.1 | - | 1 | 1291 | |||
AK122922.1 | - | 11 | 1830 |
Gene Symbol: KANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097023.1 | - | 1 | 1500 |
Gene Symbol: LINC01595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AV687190.1 | - | 1 | 70 |
Gene Symbol: LOC100131048 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125976.1 | - | 1 | 818 |
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 1970 | |||
NR_110066.1 | 3 | 1652 | ||||
NR_110067.1 | 4 | 1908 | ||||
NR_110068.1 | 2 | 1608 | ||||
NR_110069.1 | 3 | 1800 | ||||
NR_110070.1 | 5 | 1759 |
Gene Symbol: LOC441239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516745.2 | XP_011515047.1 | 3 | 1819 |
Gene Symbol: LRRC37A4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002940.2 | 6 | 6962 |
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712970.3 | XP_006713033.1 | 6 | 4965 | ||
XM_017005718.1 | XP_016861207.1 | 6 | 4965 | |||
XM_017005722.1 | XP_016861211.1 | 5 | 4947 | |||
GenBank mRNA | AF131805.1 | - | 1 | 484 |
Gene Symbol: MIEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304564.1 | NP_001291493.1 | 7 | 3011 | ||
NM_019008.5 | NP_061881.2 | 6 | 4066 | |||
NR_130789.1 | 6 | 3980 | ||||
XM_011530236.1 | XP_011528538.1 | 8 | 4134 | |||
XM_011530237.1 | XP_011528539.1 | 7 | 4254 | |||
XM_011530238.1 | XP_011528540.1 | 7 | 4338 | |||
XM_017028838.1 | XP_016884327.1 | 7 | 4232 |
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ774051.1 | - | 1 | 113 |
Gene Symbol: TLR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016562.3 | NP_057646.1 | 3 | 3776 | ||
GenBank mRNA | AF240467.1 | - | 3 | 3772 |
Gene Symbol: TSIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003255.2 | 1 | 3401 |
Gene Symbol: VPS45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000164.1 | XP_016855653.1 | 14 | 2001 |
Gene Symbol: WWC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161661.1 | NP_001155133.1 | 23 | 5016 | ||
NM_001161662.1 | NP_001155134.1 | 23 | 5013 | |||
NM_015238.2 | NP_056053.1 | 23 | 4998 |
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024646.2 | NP_078922.1 | 14 | 4000 | ||
XM_006710898.3 | XP_006710961.1 | 14 | 3887 | |||
GenBank mRNA | BX647643.1 | - | 12 | 3173 |
Set Membership: |
G-protein coupled receptor |