Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005514.7 | NP_005505.2 | 3 | 509 | ||
XM_011514557.1 | XP_011512859.1 | 3 | 432 | |||
GenBank mRNA | AF016641.1 | - | 3 | 422 | ||
AF189017.1 | - | 3 | 422 | |||
AF436098.1 | - | 3 | 422 | |||
AJ002675.1 | - | 2 | 349 | |||
AJ223602.1 | - | 2 | 349 | |||
AJ535113.1 | - | 3 | 426 | |||
AK300685.1 | - | 4 | 352 | |||
AK301008.1 | - | 3 | 329 | |||
AK310586.1 | - | 4 | 362 | |||
AK313911.1 | - | 3 | 443 | |||
BC013187.1 | - | 3 | 431 | 4344323 | ||
DC345094.1 | - | 3 | 476 | |||
FR875290.1 | - | 2 | 350 | |||
HG514457.1 | - | 3 | 422 | |||
HM989017.1 | - | 2 | 349 | |||
KU204955.1 | - | 1 | 79 | |||
L07950.1 | - | 3 | 422 | |||
L11570.1 | - | 3 | 422 | |||
L47338.1 | - | 2 | 349 | |||
M16102.1 | - | 3 | 429 | |||
M27540.1 | - | 2 | 236 | |||
M32317.1 | - | 3 | 422 | |||
U04245.1 | - | 3 | 422 | |||
U11262.1 | - | 3 | 422 | |||
U11264.1 | - | 3 | 422 | |||
U11269.1 | - | 3 | 422 | |||
U21053.1 | - | 3 | 422 | |||
U29057.1 | - | 3 | 422 | |||
U35734.1 | - | 3 | 422 | |||
X64454.1 | - | 3 | 422 | |||
X73578.1 | - | 3 | 422 | |||
Y13567.1 | - | 3 | 422 |
Gene Symbol: HLA-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008457.1 | - | 4 | 432 | 4249847 | |
D50852.1 | - | 3 | 422 | |||
D50853.1 | - | 4 | 422 | |||
D64150.1 | - | 4 | 422 | |||
FN662479.1 | - | 2 | 349 | |||
HG974570.1 | - | 4 | 422 | |||
M26429.1 | - | 3 | 422 | |||
M84171.1 | - | 3 | 422 | |||
M99389.1 | - | 4 | 422 | |||
M99390.1 | - | 4 | 422 | |||
Z46809.1 | - | 3 | 422 |