Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006827.5 | NP_006818.3 | 5 | 3405 | ||
GenBank mRNA | AK023615.1 | - | 1 | 1206 | ||
AK056156.1 | - | 1 | 1850 | |||
AL832012.2 | - | 5 | 3350 | |||
CN420777.1 | - | 1 | 270 | |||
L40391.1 | - | 1 | 710 |
Gene Symbol: LOC399715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040079.2 | 6 | 2417 | |||
GenBank mRNA | AK128185.1 | - | 7 | 2535 |