Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207067.1 | NP_001193996.1 | 11 | 1588 | ||
NM_001207069.1 | NP_001193998.1 | 11 | 1268 | |||
NM_001321688.1 | NP_001308617.1 | 11 | 1168 | |||
NM_001321690.1 | NP_001308619.1 | 11 | 1282 | |||
NM_001321691.1 | NP_001308620.1 | 11 | 1110 | |||
NM_001321693.1 | NP_001308622.1 | 10 | 1035 | |||
NM_001321694.1 | NP_001308623.1 | 11 | 1143 | |||
NM_014670.3 | NP_055485.2 | 11 | 1390 | |||
XM_017005393.1 | XP_016860882.1 | 8 | 881 | |||
GenBank mRNA | AK096489.1 | - | 10 | 1145 | ||
AK223245.1 | - | 11 | 1197 | |||
AK297227.1 | - | 11 | 1281 | |||
AK301511.1 | - | 11 | 1268 | |||
AL833518.1 | - | 11 | 1544 | |||
BC001804.1 | - | 11 | 1012 | 3354883 | ||
BC026303.1 | - | 11 | 1197 | 4284614 | ||
D13630.1 | - | 11 | 1213 |