Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002124.3 | NP_002115.2 | 2 | 225 | ||
GenBank mRNA | AF010142.1 | - | 1 | 44 | ||
AK291987.1 | - | 2 | 225 | |||
AY961072.1 | - | 2 | 131 | |||
AY961073.1 | - | 2 | 131 | |||
BC033827.1 | - | 2 | 174 | 5216243 | ||
BC108922.1 | - | 2 | 186 | 40023604 | ||
HG974574.1 | - | 2 | 131 | |||
HG974596.1 | - | 2 | 131 | |||
HG974600.1 | - | 2 | 131 | |||
HM067861.1 | - | 2 | 131 | |||
HM067862.1 | - | 2 | 131 | |||
HM067863.1 | - | 2 | 131 | |||
KJ534866.1 | - | 2 | 131 | |||
L02545.1 | - | 2 | 145 | |||
M16957.1 | - | 1 | 44 | |||
M16958.1 | - | 1 | 44 | |||
M16959.1 | - | 1 | 44 | |||
M17378.1 | - | 1 | 44 | |||
M20430.1 | - | 2 | 174 | |||
M20504.1 | - | 2 | 166 | |||
M28583.1 | - | 2 | 170 | |||
M28584.1 | - | 2 | 102 | |||
M30179.1 | - | 2 | 131 |
Gene Symbol: HLA-DRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M30180.1 | - | 2 | 102 |