Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 3441 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 2507 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3308 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 2995 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 3570 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 3834 | |||
NM_003879.5 | NP_003870.4 | 10 | 3413 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4308 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3277 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3356 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4243 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6260 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4116 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 3454 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3079 |