Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 2 | 226 | ||
NR_111964.1 | 2 | 418 | ||||
GenBank mRNA | AV739733.1 | - | 2 | 270 | ||
BC027888.1 | - | 2 | 214 | 5222247 | ||
BC107710.1 | - | 2 | 230 | 6699133 | ||
BC146914.1 | - | 2 | 177 | 9056842 | ||
X52149.1 | - | 2 | 404 |
Gene Symbol: CCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002983.2 | NP_002974.1 | 2 | 240 | ||
GenBank mRNA | AF043339.1 | - | 1 | 72 | ||
BC071834.1 | - | 2 | 212 | 5935287 | ||
BC171831.1 | - | 2 | 174 | 9054485 | ||
BC171891.1 | - | 2 | 174 | 9054545 | ||
BX103455.1 | - | 2 | 240 | |||
D00044.1 | - | 2 | 221 | |||
M23452.1 | - | 2 | 221 | |||
M25315.1 | - | 2 | 221 | |||
X03754.1 | - | 2 | 221 |
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 2 | 216 | ||
GenBank mRNA | AA380745.1 | - | 2 | 216 | ||
AI302038.1 | - | 2 | 117 | |||
BC007783.2 | - | 2 | 211 | 4308472 | ||
BC146919.1 | - | 2 | 177 | 9056847 | ||
BC147008.1 | - | 2 | 177 | 9056936 | ||
BC147013.1 | - | 2 | 177 | 9056941 | ||
BT006783.1 | - | 2 | 141 | |||
CF128666.1 | - | 2 | 195 |