Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526449.2 | XP_011524751.1 | 11 | 1360 |
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025700.1 | - | 7 | 1485 |
Gene Symbol: AGPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358704.1 | - | 2802 |
Gene Symbol: ANKRD20A11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043199.1 | - | 2 | 948 |
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721447.2 | XP_006721510.2 | 1 | 1937 | ||
XM_017024182.1 | XP_016879671.1 | 1 | 1937 |
Gene Symbol: DCAF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005248169.3 | XP_005248226.1 | 3 | 6799 | ||
XM_005248171.3 | XP_005248228.1 | 2 | 6695 | |||
XM_006713966.3 | XP_006714029.1 | 4 | 6970 | |||
XM_017008327.1 | XP_016863816.1 | 2 | 8264 | |||
XM_017008328.1 | XP_016863817.1 | 2 | 8249 |
Gene Symbol: ERN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU608100.1 | - | 1 | 396 |
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127774.1 | - | 3 | 1881 |
Gene Symbol: FANCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021922.2 | NP_068741.1 | 10 | 2234 | ||
XM_005248885.2 | XP_005248942.1 | 10 | 2235 | |||
XM_005248886.2 | XP_005248943.1 | 9 | 2187 | |||
XM_005248887.2 | XP_005248944.1 | 9 | 2130 | |||
XM_005248888.3 | XP_005248945.1 | 8 | 2063 | |||
XM_011514343.2 | XP_011512645.1 | 10 | 1949 | |||
GenBank mRNA | AF265210.1 | - | 10 | 2234 | ||
BC046359.1 | - | 10 | 2240 | 6154799 |
Gene Symbol: LINC00581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103790.1 | 1 | 45 | |||
GenBank mRNA | BX118339.1 | - | 1 | 49 |
Gene Symbol: LOC101928322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120637.1 | 4 | 492 | |||
GenBank mRNA | DA490058.1 | - | 4 | 491 |
Gene Symbol: LOC400576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127731.1 | - | 2 | 2514 |
Gene Symbol: LRRC37A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126994.1 | - | 1 | 448 |
Gene Symbol: MYH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749516.1 | - | 5 | 2982 |
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125214.1 | - | 2 | 927 |
Gene Symbol: NPRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125420.1 | - | 1 | 220 |
Gene Symbol: NUDT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018283.3 | NP_060753.1 | 3 | 1696 | ||
GenBank mRNA | AK001818.1 | - | 3 | 1696 | ||
BC064607.1 | - | 3 | 1515 | |||
BC107875.1 | - | 3 | 1569 | 6451308 | ||
BC133015.1 | - | 3 | 1637 | 40147037 | ||
BC133017.1 | - | 3 | 1638 | 40147039 |
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648172.1 | - | 1 | 2439 |
Gene Symbol: PDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020786.2 | NP_065837.1 | 2 | 5458 | ||
XM_005256064.4 | XP_005256121.1 | 2 | 5505 | |||
XM_005256066.4 | XP_005256123.1 | 2 | 5547 | |||
XM_005256067.4 | XP_005256124.1 | 2 | 5572 | |||
XM_005256068.4 | XP_005256125.1 | 3 | 5519 | |||
XM_017023506.1 | XP_016878995.1 | 3 | 5653 |
Gene Symbol: PLAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182832.2 | NP_878252.2 | 1 | 6581 | ||
GenBank mRNA | AF269287.1 | - | 1 | 2830 | ||
AK027868.1 | - | 1 | 4243 | |||
L13197.1 | - | 2 | 2213 |
Gene Symbol: RGS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008530.1 | XP_016864019.1 | 4 | 4262 |
Gene Symbol: S100PBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647667.1 | - | 7 | 2768 |
Gene Symbol: SAFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749251.1 | - | 1 | 841 |
Gene Symbol: SAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537502.1 | - | 7 | 1781 |
Gene Symbol: SLC1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209199.1 | - | 1 | 1936 |
Gene Symbol: SLC2A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030777.3 | NP_110404.1 | 5 | 2394 | ||
XM_011529060.2 | XP_011527362.1 | 5 | 2477 | |||
XM_011529061.2 | XP_011527363.1 | 5 | 4034 | |||
XM_011529062.2 | XP_011527364.1 | 4 | 2401 | |||
XM_011529065.2 | XP_011527367.1 | 4 | 2348 | |||
XM_017028087.1 | XP_016883576.1 | 4 | 2251 | |||
GenBank mRNA | AF248053.1 | - | 5 | 2546 | ||
AF321240.1 | - | 5 | 2349 | |||
AK055548.1 | - | 5 | 2394 | |||
AL137188.3 | - | 4 | 2305 |
Gene Symbol: SLC30A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013309.5 | NP_037441.2 | 8 | 4560 | ||
XM_011521997.2 | XP_011520299.1 | 8 | 5064 | |||
XM_017022560.1 | XP_016878049.1 | 7 | 4366 |
Gene Symbol: SSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292008.1 | NP_001278937.1 | 8 | 5252 | ||
NM_003144.4 | NP_003135.2 | 8 | 5456 | |||
NR_120448.1 | 7 | 5377 | ||||
GenBank mRNA | AK024294.1 | - | 1 | 791 | ||
AK025753.1 | - | 1 | 1544 |
Gene Symbol: TNFRSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001561.5 | NP_001552.2 | 9 | 4031 | ||
XM_011541386.2 | XP_011539688.1 | 8 | 4046 | |||
GenBank mRNA | AK125490.1 | - | 1 | 2049 |
Gene Symbol: TRAF3IP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043575.1 | - | 2 | 511 |
Gene Symbol: ZC3HAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024625.3 | NP_078901.3 | 9 | 2892 | ||
GenBank mRNA | AF138863.1 | - | 6 | 1202 | ||
BC002935.1 | - | 6 | 1604 | |||
BC025308.1 | - | 9 | 2823 | 5418915 | ||
BC033105.1 | - | 9 | 2823 | 5418956 | ||
BC040956.1 | - | 9 | 2889 | 5531412 |
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531635.2 | XP_011529937.1 | 1 | 2016 | ||
XM_017007769.1 | XP_016863258.1 | 1 | 2016 |