Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006703.3 | NP_006694.1 | 2 | 484 | ||
GenBank mRNA | AF062529.1 | - | 2 | 319 | ||
AF062530.1 | - | 2 | 319 | |||
AK313440.1 | - | 2 | 387 | |||
BC007727.2 | - | 2 | 445 | 4303483 | ||
BG714472.1 | - | 2 | 483 | |||
BT019984.1 | - | 2 | 166 | |||
BT019985.1 | - | 2 | 166 |
Gene Symbol: NUDT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304963.1 | NP_001291892.1 | 1 | 320 | ||
NM_153183.3 | NP_694853.1 | 2 | 383 | |||
GenBank mRNA | AF469196.1 | - | 2 | 385 | ||
AK291952.1 | - | 1 | 300 | |||
BC049383.1 | - | 1 | 241 | 5725355 | ||
BC050700.1 | - | 1 | 299 | 5723622 | ||
BX449265.2 | - | 1 | 320 |
Gene Symbol: NUDT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018159.3 | NP_060629.2 | 1 | 326 | ||
GenBank mRNA | AK001490.1 | - | 1 | 213 | ||
BC009942.2 | - | 1 | 280 | 4303422 | ||
BX419815.2 | - | 1 | 312 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 6 | 617 | ||
GenBank mRNA | BU521288.1 | - | 6 | 540 |