siRNA Details
Gene Symbol: CYCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018947.5 | NP_061820.1 | 3 | 472 | ||
GenBank mRNA | AK311836.1 | - | 3 | 403 | ||
AL713681.1 | - | 3 | 362 | |||
BC005299.1 | - | 3 | 369 | 3838954 | ||
BC008475.1 | - | 3 | 371 | 4284529 | ||
BC008477.1 | - | 3 | 371 | 4290897 | ||
BC009578.1 | - | 3 | 371 | 3961416 | ||
BC009579.1 | - | 3 | 371 | 4045034 | ||
BC009582.1 | - | 3 | 371 | 4082112 | ||
BC009587.1 | - | 3 | 371 | 4096763 | ||
BC009602.1 | - | 3 | 371 | 4134675 | ||
BC009607.1 | - | 3 | 375 | 4287362 | ||
BC014359.1 | - | 3 | 375 | 3930323 | ||
BC014361.1 | - | 3 | 375 | 3931721 | ||
BC015130.1 | - | 3 | 375 | 3932265 | ||
BC016006.1 | - | 3 | 375 | 4656402 | ||
BC021994.1 | - | 3 | 375 | 4096609 | ||
BC022330.1 | - | 3 | 371 | 4280695 | ||
BC024216.1 | - | 3 | 414 | |||
BC067222.1 | - | 3 | 375 | 3685258 | ||
BC068464.1 | - | 3 | 371 | 4826114 | ||
BC070156.1 | - | 3 | 371 | 6451982 | ||
BC070346.1 | - | 3 | 372 | 3826017 | ||
BC071761.1 | - | 3 | 371 | 4656503 | ||
BT006946.1 | - | 2 | 302 | |||
BU597562.1 | - | 3 | 450 | |||
CR542175.1 | - | 2 | 302 | |||
D00265.1 | - | 1 | 71 |
Gene Symbol: CYCSP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001560.1 | 1 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
