Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECSIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142464.2 | NP_001135936.1 | 7 | 1342 | ||
NM_001142465.2 | NP_001135937.1 | 6 | 849 | |||
NM_001243204.1 | NP_001230133.1 | 8 | 1380 | |||
NM_016581.4 | NP_057665.2 | 8 | 1491 | |||
GenBank mRNA | AF243044.1 | - | 7 | 1390 | ||
BC000193.1 | - | 8 | 1439 | 3352568 | ||
BC005119.1 | - | 8 | 1434 | 2819610 | ||
BC008279.1 | - | 7 | 1307 | 3639801 | ||
BU186092.1 | - | 5 | 622 |