Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000146.3 | NP_000137.2 | 4 | 822 | ||
GenBank mRNA | AF147331.1 | - | 3 | 433 | ||
AK026534.1 | - | 1 | 822 | |||
AK130191.1 | - | 4 | 822 | |||
AK130205.1 | - | 4 | 817 | |||
AK131048.1 | - | 4 | 822 | |||
AK131050.1 | - | 4 | 822 | |||
AK131053.1 | - | 4 | 836 | |||
AK307065.1 | - | 4 | 812 | |||
AY207005.1 | - | 4 | 826 | |||
BC002991.1 | - | 4 | 790 | 2905327 | ||
BC004245.1 | - | 4 | 811 | 3615360 | ||
BC008439.1 | - | 1 | 822 | 4093482 | ||
BC008441.1 | - | 1 | 822 | |||
BC013928.1 | - | 1 | 826 | 3839120 | ||
BC016346.1 | - | 1 | 826 | 4066010 | ||
BC016354.1 | - | 4 | 826 | 4092360 | ||
BC016715.1 | - | 1 | 826 | 4067055 | ||
BC018990.2 | - | 1 | 794 | 3503710 | ||
BC021670.1 | - | 1 | 825 | 4051089 | ||
BC058820.1 | - | 1 | 815 | 4049525 | ||
BC062708.1 | - | 1 | 825 | 4774403 | ||
BX571748.1 | - | 1 | 821 | |||
M10119.1 | - | 4 | 702 | |||
M11147.1 | - | 4 | 774 | |||
M12938.1 | - | 3 | 525 |