Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033323.3 | 8 | 958 | |||
GenBank mRNA | AF416611.1 | - | 8 | 915 | ||
AF420250.1 | - | 8 | 915 | |||
AK021688.1 | - | 8 | 915 | |||
AK292107.1 | - | 4 | 391 | |||
AK303622.1 | - | 3 | 737 | |||
AK315927.1 | - | 3 | 571 | |||
AL117583.1 | - | 1 | 703 | |||
AL833016.1 | - | 7 | 1601 | |||
BC007488.2 | - | 8 | 1012 | |||
BC082753.1 | - | 4 | 371 | 4542435 | ||
BC106049.1 | - | 4 | 439 |