Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013302.3 | NP_037434.1 | 18 | 6818 | ||
XM_017023193.1 | XP_016878682.1 | 18 | 6529 | |||
XM_017023194.1 | XP_016878683.1 | 17 | 6762 |