Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 3 | 472 | ||
NM_182931.2 | NP_891847.1 | 4 | 620 | |||
XM_005250493.1 | XP_005250550.1 | 3 | 539 | |||
XM_011516400.1 | XP_011514702.1 | 5 | 746 | |||
XM_017012435.1 | XP_016867924.1 | 3 | 531 | |||
GenBank mRNA | AF519459.1 | - | 3 | 287 | ||
AK000940.1 | - | 3 | 287 | |||
AY147037.1 | - | 4 | 620 | |||
AY157990.1 | - | 2 | 86 | |||
BC001296.2 | - | 4 | 582 | |||
BC040004.1 | - | 3 | 490 | |||
BC053906.1 | - | 3 | 528 | |||
BC062583.1 | - | 4 | 592 | |||
BC142987.1 | - | 3 | 502 | |||
JQ809698.1 | - | 2 | 86 |