Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORCS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145783.1 | NP_001139255.1 | 2 | 590 | ||
NM_001145784.1 | NP_001139256.1 | 2 | 590 | |||
GenBank mRNA | BC000489.2 | - | 2 | 139 | ||
BC004449.1 | - | 2 | 139 | |||
BC010178.2 | - | 2 | 148 | |||
BC010931.1 | - | 2 | 155 | |||
BC157837.1 | - | 2 | 421 | |||
BC157868.1 | - | 2 | 203 | 9057087 | ||
BC171829.1 | - | 2 | 421 | |||
BC171869.1 | - | 2 | 203 |
Gene Symbol: BORCS8-MEF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027307.1 | 2 | 590 | |||
NR_027308.1 | 2 | 590 | ||||
GenBank mRNA | AK301086.1 | - | 2 | 157 | ||
AK316328.1 | - | 2 | 157 | |||
X63380.1 | - | 2 | 127 |