Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORCS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145783.1 | NP_001139255.1 | 4 | 769 | ||
NM_001145784.1 | NP_001139256.1 | 4 | 769 | |||
GenBank mRNA | BC000489.2 | - | 4 | 318 | ||
BC004449.1 | - | 4 | 318 | |||
BC010178.2 | - | 4 | 327 | |||
BC010931.1 | - | 4 | 334 | |||
BC157837.1 | - | 4 | 600 | |||
BC157868.1 | - | 4 | 382 | 9057087 | ||
BC171829.1 | - | 4 | 600 | |||
BC171869.1 | - | 4 | 382 |
Gene Symbol: BORCS8-MEF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005919.3 | NP_005910.1 | 2 | 591 | ||
NR_027307.1 | 4 | 769 | ||||
NR_027308.1 | 4 | 769 | ||||
GenBank mRNA | AK301086.1 | - | 4 | 336 | ||
AK316328.1 | - | 4 | 336 | |||
BC126245.1 | - | 1 | 14 | 8991961 | ||
BC136457.1 | - | 1 | 14 | 9020446 | ||
BC171767.1 | - | 1 | 14 | 9054421 | ||
X63380.1 | - | 4 | 306 | |||
X68502.1 | - | 2 | 113 |